Skip to Content
Merck
All Photos(1)

Documents

39931

Sigma-Aldrich

1-Butyl-3-methylimidazolium tetrafluoroborate

for catalysis, ≥98.5% (HPLC)

Synonym(s):

BMIMBF4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15BF4N2
CAS Number:
Molecular Weight:
226.02
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

for catalysis

Quality Level

Assay

≥98.5% (HPLC)

impurities

≤0.05% water

refractive index

n20/D 1.52

density

1.21 g/mL at 20 °C (lit.)

anion traces

bromide (Br-): ≤25 mg/kg
chloride (Cl-): ≤25 mg/kg
nitrate (NO3-): ≤25 mg/kg
phosphate (PO43-): ≤25 mg/kg
sulfate (SO42-): ≤10 mg/kg

SMILES string

F[B-](F)(F)F.CCCCn1cc[n+](C)c1

InChI

1S/C8H15N2.BF4/c1-3-4-5-10-7-6-9(2)8-10;2-1(3,4)5/h6-8H,3-5H2,1-2H3;/q+1;-1

InChI key

LSBXQLQATZTAPE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Butyl-3-methylimidazolium tetrafluoroborate is a high-purity ionic liquid (HPIL) with low water and halogen content.

Application

1-Butyl-3-methylimidazolium tetrafluoroborate has been used as a solvent for the synthesis of zinc sulfide (ZnS) nanoparticles in the absence stabilizing agents or capping agents. It can also be used as an extractant for the removal of dibenzothiophene (DBS) from liquid fuels via extractive desulfurization.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

550.4 °F

Flash Point(C)

288 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

ZnS nanoparticle synthesis in 1-butyl-3-methylimidazolium tetrafluoroborate by simple heating.
Kareem TA and Kaliani AA.
Arabian Journal of Chemistry (2015)
Preparation of 1?Butyl?3?Methyl Imidazolium?Based Room Temperature Ionic Liquids
Dupont, Jairton, et al.
Organic Syntheses, 236-236 (2003)
Solubility of carbon dioxide, ethane, methane, oxygen, nitrogen, hydrogen, argon, and carbon monoxide in 1-butyl-3-methylimidazolium tetrafluoroborate between temperatures 283K and 343K and at pressures close to atmospheric
Jacquemin, Johan, et al.
The Journal of Chemical Thermodynamics, 38.4, 490-502 (2006)
Synthesis, characterization and application of 1-butyl-3-methylimidazolium tetrafluoroborate for extractive desulfurization of liquid fuel.
Dharaskar SA, et al.
Arabian Journal of Chemistry, 9(4), 578-587 (2016)
Electro?induced Cationic Polymerization of Vinyl Ethers by Using Ionic Liquid 1?Butyl?3?methylimidazolium Tetrafluoroborate as Initiator
Yang, Feng, et al.
Macromolecular Chemistry and Physics, 216.4, 380-385 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service