Skip to Content
Merck
All Photos(2)

Documents

38332

Supelco

Scopoletin

analytical standard

Synonym(s):

6-Methoxyumbelliferone, 6-Methylesculetin, 7-Hydroxy-6-methoxy-2H-1-benzopyran-2-one, 7-Hydroxy-6-methoxycoumarin, Buxuletin, Chrysatropic acid, Escopoletin, Esculetin-6-methyl ether, Gelseminic acid, Murrayetin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8O4
CAS Number:
Molecular Weight:
192.17
Beilstein:
156296
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥97.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

203-205 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc2C=CC(=O)Oc2cc1O

InChI

1S/C10H8O4/c1-13-9-4-6-2-3-10(12)14-8(6)5-7(9)11/h2-5,11H,1H3

InChI key

RODXRVNMMDRFIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Scopoletin is a coumarin compound and pharmacologically active chemical that is found in various plant species.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Scopoletin may be used as a reference standard for the determination of scopoletin in:
  • Roots, stems, branches, and seeds as well as leaves of Chimonanthus nitens Oliv. by ultra-high performance liquid chromatography/tandem quadrupole-time-of-flight mass spectrometry (UHPLC-QTOF-MS/MS) equipped with modified mass deflect filter or diagnostic product ions/neutral loss filter.
  • Canscora decussate (South Indian Shankhpushpi) extracts by high performance thin layer chromatography (HPTLC).
  • Morinda tinctoria Roxb. leaf by HPTLC in conjunction with HPLC and gas chromatography with MS.

Biochem/physiol Actions

Scopoletin significantly increases activity of lipoprotein lipase in adipocytes by promoting transcription of the LPL gene. It also partially reverses inhibition of LPL by TNF-α.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

This compound is commonly found in plants of the genus: achillea angelica glycyrrhiza lycium pimpinella

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comprehensive profiling and characterization of coumarins from roots, stems, leaves, branches, and seeds of Chimonanthus nitens Oliv. using ultra-performance liquid chromatography/quadrupole-time-of-flight mass spectrometry combined with modified mass defect filter.
Tan T, et al.
Journal of Pharmaceutical and Biomedical Analysis, 141(2-3), 140-148 (2017)
Structural characterization and discrimination of Chimonanthus nitens Oliv. leaf from different geographical origins based on multiple chromatographic analysis combined with chemometric methods.
Huang WP, et al.
Journal of Pharmaceutical and Biomedical Analysis, 154(2-3), 236-244 (2018)
HPTLC Finger print analysis and Phytochemical Investigation of Morinda tinctoria Roxb. Leaf extracts by HPLC and GS MS.
Arunachalam KD, et al.
International Journal of Pharmacy and Pharmaceutical Sciences, 7(2), 360-366 (2015)
Fazeelat Karamat et al.
BMC plant biology, 12, 152-152 (2012-08-31)
Furanocoumarins are molecules with proven therapeutic properties and are produced in only a small number of medicinal plant species such as Ruta graveolens. In vivo, these molecules play a protective role against phytophageous insect attack. Furanocoumarins are members of the
M Baz et al.
Journal of applied microbiology, 112(4), 782-792 (2012-02-02)
The early molecular events underlying the elicitation of plant defence reactions by Gram-positive bacteria are relatively unknown. In plants, calcium and reactive oxygen species are commonly involved as cellular messengers of a wide range of biotic stimuli from pathogenic to

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service