Skip to Content
Merck
All Photos(1)

Key Documents

37905

Supelco

Thiacloprid

PESTANAL®, analytical standard

Synonym(s):

[3-(6-Chloro-3-pyridinylmethyl)-2-thiazolidinylidene]cyanamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9ClN4S
CAS Number:
Molecular Weight:
252.72
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Clc1ccc(CN2CCS\C2=N\C#N)cn1

InChI

1S/C10H9ClN4S/c11-9-2-1-8(5-13-9)6-15-3-4-16-10(15)14-7-12/h1-2,5H,3-4,6H2/b14-10+

InChI key

HOKKPVIRMVDYPB-GXDHUFHOSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Thiacloprid is a chloronicotinyl insecticide, which can be commonly used in agriculture in order to control various sucking insects like aphids, whiteflies and some jassids. It can also be actively used against weevils, leaf miners and various species of beetles.

Find more information here: Neonicotinoids

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Repr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jochen P Zubrod et al.
Environmental science & technology, 51(16), 9280-9287 (2017-07-07)
Disturbance regimes determine communities' structure and functioning. Nonetheless, little effort has been undertaken to understand interactions of press and pulse disturbances. In this context, leaf-shredding macroinvertebrates can be chronically exposed to wastewater treatment plant effluents (i.e., press disturbance) before experiencing
Léa Tison et al.
The Journal of experimental biology, 220(Pt 20), 3695-3705 (2017-08-19)
Learning and memory play a central role in the behavior and communication of foraging bees. We have previously shown that chronic uptake of the neonicotinoid thiacloprid affects the behavior of honey bees in the field. Foraging behavior, homing success, navigation
Makoto Ihara et al.
British journal of pharmacology, 175(11), 1999-2012 (2017-06-16)
Neonicotinoid insecticides interact with the orthosteric site formed at subunit interfaces of insect nicotinic ACh (nACh) receptors. However, their interactions with the orthosteric sites at α-non α and α-α subunit interfaces remain poorly understood. The aim of this study was
Céline Lukowicz et al.
Environmental health perspectives, 126(6), 067007-067007 (2018-06-29)
Epidemiological evidence suggests a link between pesticide exposure and the development of metabolic diseases. However, most experimental studies have evaluated the metabolic effects of pesticides using individual molecules, often at nonrelevant doses or in combination with other risk factors such
Cornelis A M van Gestel et al.
Ecotoxicology (London, England), 26(3), 320-328 (2017-01-25)
In a recent study, we showed that the springtail Folsomia candida was quite sensitive the neonicotinoid insecticides imidacloprid and thiacloprid. This study aimed at determining the toxicity of both compounds to F. candida following exposure over three generations, in natural

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service