Skip to Content
Merck
All Photos(1)

Key Documents

37018

Supelco

Tenuazonic acid

analytical standard

Synonym(s):

(5S)-3-Acetyl-1,5-dihydro-4-hydroxy-5-[(1S)-1-methylpropyl]-2H-pyrrol-2-one, (S)-3-Acetyl-5-(S)-sec-butyltetramic acid, (S)-3-Acetyl-5-sec-butyl-4-hydroxy-3-pyrrolin-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H15NO3
CAS Number:
Molecular Weight:
197.23
Beilstein:
3589518
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CC[C@H](C)[C@@H]1NC(=O)C(C(C)=O)=C1O

InChI

1S/C10H15NO3/c1-4-5(2)8-9(13)7(6(3)12)10(14)11-8/h5,8,13H,4H2,1-3H3,(H,11,14)/t5-,8-/m0/s1

InChI key

CEIZFXOZIQNICU-XNCJUZBTSA-N

General description

Tenuazonic acid is a biologically active compound and its mode of action involves the inhibition of protein biosynthesis.
Tenuazonic acid is a mycotoxin, which belongs to the class of tetramic acids, the largest family of natural products. It is generally isolated from phytopathogenic fungal species like Alternaria alternata, A. longipes and A. tenuissima.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tenuazonic acid may be used as an analytical reference standard for the quantification of the analyte in the following:
  • Cornflakes using high-performance liquid chromatography (HPLC) technique.
  • Human urine samples using isotope dilution assay method and liquid chromatography coupled to a hybrid triple quadrupole/linear ion trap mass spectrometer.
  • Cereals using high-performance liquid chromatography–electrospray ionization ion-trap multistage mass spectrometry (HPLC- ESI ion-trap (IT) MS2). ESI Fourier transform-ion cyclotron resonance tandem mass spectrometry (FTICR-MS2) technique is used for product characterization, post derivatization with 2,4-dinitrophenylhydrazine.

Reconstitution

Dried down, ~100 μg/mL after reconstitution with 1 mL of water

Analysis Note

purity : ≥96.0% (HPLC)

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Recent advances in tenuazonic acid as a potential herbicide
Chen S and Qiang S
Pesticide Biochemistry and Physiology (2017)
Simultaneous determination of ochratoxin A and cyclopiazonic, mycophenolic, and tenuazonic acids in cornflakes by solid-phase microextraction coupled to high-performance liquid chromatography
Aresta A, et al.
Journal of Agricultural and Food Chemistry, 51(18), 5232-5237 (2003)
Determination of tenuazonic acid in human urine by means of a stable isotope dilution assay
Asam S, et al.
Analytical and Bioanalytical Chemistry, 405(12), 4149-4158 (2013)
Andrea Patriarca et al.
International journal of food microbiology, 291, 135-143 (2018-12-01)
The group of the small-spored Alternaria species is particularly relevant in foods due to its high frequency and wide distribution in different crops. These species are responsible for the accumulation of mycotoxins and bioactive secondary metabolites in food. The taxonomy
Dominik Braun et al.
Frontiers in chemistry, 8, 423-423 (2020-06-09)
Exposure to natural food contaminants during infancy may influence health consequences later in life. Hence, breast milk may serve as a vehicle to transport these contaminants, including mycotoxins, from mothers to their infants. Analytical methods mostly focused on single exposures

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service