Skip to Content
Merck

Skip To

36979

Polynuclear aromatic hydrocarbons 16 solution

analytical standard, in acetonitrile

Synonym(s):

PAH 16 Standard Solution

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

description

suitable for EPA Method 610

quality

in acetonitrile

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Polynuclear aromatic hydrocarbons 16 solution has been used as a standard in the estimation of polynuclear aromatic hydrocarbons, carbon and heavy metals, which are present in street dust, using high performance liquid chromatography (HPLC).

Other Notes

10 μg each/ml acenaphthene, acenaphthylene, anthracene, benzo[a]anthracene, benzo[b]fluoranthene, benzo[k]fluoranthene, benzo[ghi]perylene, benzo[a]pyrene, chrysene, dibenzo[a,h]anthracene, fluoranthene, fluorene, indeno[1,2,3-cd]pyrene, naphthalene, phenanthrene and pyrene

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
36991CRM4890532273
technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

format

multi-component solution

format

multi-component solution

format

multi-component solution

format

single component solution

Quality Level

100

Quality Level

100

Quality Level

300

Quality Level

100

grade

analytical standard

grade

analytical standard

grade

certified reference material, TraceCERT®

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

limited shelf life, expiry date on the label

quality

in acetonitrile

quality

in cyclohexane

quality

-

quality

-


Still not finding the right product?


signalword

Danger

flash_point_f

35.6 °F

flash_point_c

2 °C

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters