Skip to Content
Merck
All Photos(1)

Documents

36746

Supelco

2-Chlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OH
CAS Number:
Molecular Weight:
128.56
Beilstein:
1905114
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.558 (lit.)

bp

175-176 °C (lit.)

mp

8 °C (lit.)

density

1.241 g/mL at 25 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1ccccc1Cl

InChI

1S/C6H5ClO/c7-5-3-1-2-4-6(5)8/h1-4,8H

InChI key

ISPYQTSUDJAMAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

147.2 °F - closed cup

Flash Point(C)

64.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thi Thu Nga Nguyen et al.
The Journal of antimicrobial chemotherapy, 70(3), 731-738 (2014-10-23)
HIV resistance to the integrase inhibitor raltegravir in treated patients is characterized by distinct resistance pathways. We hypothesize that differences in the in vivo dynamics of HIV resistance to raltegravir are due to the genetic context of the integrase present
Esther Martí et al.
The Science of the total environment, 409(14), 2707-2716 (2011-05-03)
Three chlorophenolic compounds (2-chlorophenol, 2,4,6-trichlorophenol, and pentachlorophenol) were tested to assess their effects on two soils with different properties: a granitic soil (Haplic Arenosol) and a calcareous one (Calcaric Regosol). Different concentrations of the pollutants (ranging from 0.001 to 10,000
Joan Caliz et al.
FEMS microbiology ecology, 78(1), 150-164 (2011-03-24)
The impact of 2-monochlorophenol (MCP), 2,4,6-trichlorophenol (TCP) and pentachlorophenol (PCP) on the microbial community of an acidic forest soil was studied under controlled laboratory conditions by spiking microcosms with the pollutants at concentrations ranging from 0.1 to 5000 mg kg(-1).
Mohammad Zain Khan et al.
Journal of hazardous materials, 190(1-3), 222-228 (2011-04-07)
2-Chlorophenol (2-CP) degrading aerobic granules were cultivated in a sequencing batch reactor (SBR) in presence of glucose. The organic loading rate (OLR) was increased from 6.9 to 9.7 kg COD m(-3)d(-1) (1150-1617 mg L(-1)COD per cycle) during the experiment. The
Julio Alonso-Padilla et al.
PLoS neglected tropical diseases, 8(12), e3259-e3259 (2014-12-05)
The discovery of new therapeutic options against Trypanosoma cruzi, the causative agent of Chagas disease, stands as a fundamental need. Currently, there are only two drugs available to treat this neglected disease, which represents a major public health problem in

Protocols

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Analytical standard separation of various phenols for research and industrial applications.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service