Skip to Content
Merck

36684

3-Aminophenol

PESTANAL®, analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
H2NC6H4OH
CAS Number:
Molecular Weight:
109.13
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
209-711-2
Beilstein/REAXYS Number:
636059
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

bp

164 °C/11 mmHg (lit.)

mp

120-124 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Nc1cccc(O)c1

InChI

1S/C6H7NO/c7-5-2-1-3-6(8)4-5/h1-4,8H,7H2

InChI key

CWLKGDAVCFYWJK-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under 3-Aminophenol


pictograms

Exclamation markEnvironment

signalword

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 2

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library