Skip to Content
Merck
All Photos(1)

Documents

36187

Supelco

Parathion-methyl

PESTANAL®, analytical standard

Synonym(s):

O,O-Dimethyl O-(4-nitrophenyl) phosphorothioate, Methyl parathion

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10NO5PS
CAS Number:
Molecular Weight:
263.21
Beilstein:
8905814
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

COP(=S)(OC)Oc1ccc(cc1)[N+]([O-])=O

InChI

1S/C8H10NO5PS/c1-12-15(16,13-2)14-8-5-3-7(4-6-8)9(10)11/h3-6H,1-2H3

InChI key

RLBIQVVOMOPOHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Parathion-methyl is an organophosphorus insecticide and acaricide, which is widely used in controlling insect pests of agricultural crops, fruits and vegetables.

Application

Parathion-methyl may be used as a reference standard in the determination of parathion-methyl in water samples using high performance liquid chromatography with electrochemical detection (HPLC-EC).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Acute Tox. 3 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

114.8 °F - open cup

Flash Point(C)

46 °C - open cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Comparative study of three immunoassays based on monoclonal antibodies for detection of the pesticide parathion-methyl in real samples.
KolosovaY A, et al.
Analytica Chimica Acta, 511(2), 323-331 (2004)
Patrick B Dennis et al.
Biomacromolecules, 13(7), 2037-2045 (2012-06-02)
Organophosphates are some of the most acutely toxic compounds synthesized on an industrial scale, and organophosphorus hydrolase (OPH) has the ability to hydrolyze and inactivate a number of these chemicals. However, OPH activity is vulnerable to harsh environmental conditions that
Dan Zhang et al.
The Analyst, 137(11), 2629-2636 (2012-04-14)
A novel composite of vinyl group functionalized multiwalled carbon nanotubes (MWCNTs) molecularly imprinted polymer (MIP) was synthesized and applied as a molecular recognition element to construct an electrochemical sensor for parathion-methyl in this paper. The special molecular recognition properties of
Yanbo Zeng et al.
Journal of hazardous materials, 217-218, 315-322 (2012-04-13)
A sensitive electrochemical differential pulse voltammetry method was developed for detecting methyl parathion based on multiwalled carbon nanotubes-poly(acrylamide) (MWCNTs-PAAM) nanocomposite film modified glassy carbon electrode. The novel MWCNTs-PAAM nanocomposite, containing high content of amide groups, was synthesized by PAAM polymerizing
Scott A Trammell et al.
Chemical communications (Cambridge, England), 48(34), 4121-4123 (2012-03-22)
Using a low power green laser, we have demonstrated a rate acceleration of ~2-fold for the hydrolysis of methyl parathion by irradiating the plasmon absorption band of Au nanoparticles capped with a Cu(bpy) catalyst.

Protocols

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Test your food for fipronil contamination using our analytical standards, certified reference materials, solvents, and columns for analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service