Skip to Content

34522

Prochloraz Metabolite BTS44596

PESTANAL®, analytical standard

Synonym(s):

N′-Formyl-N-propyl-N-[2-(2,4,6-trichlorophenoxy)ethyl]urea

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C13H15Cl3N2O3
CAS Number:
Molecular Weight:
353.63
UNSPSC Code:
77101502
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
9142204
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

format

neat

SMILES string

ClC1=C(OCCN(CCC)C(NC([H])=O)=O)C(Cl)=CC(Cl)=C1

InChI

1S/C13H15Cl3N2O3/c1-2-3-18(13(20)17-8-19)4-5-21-12-10(15)6-9(14)7-11(12)16/h6-8H,2-5H2,1H3,(H,17,19,20)

InChI key

RHDVQZWCBQXOJW-UHFFFAOYSA-N

Application

  • Dissipation and Dietary Risk Assessment of Prochloraz in Strawberries under Greenhouse Conditions.: This research investigates the dissipation rates and dietary risks of Prochloraz in strawberries, emphasizing the role of its metabolite BTS44596 in assessing the safety and environmental impact of fungicide applications in agriculture (Wang et al., 2023).
  • The journey of prochloraz pesticide in Citrus sinensis: Residual distribution, impact on transcriptomic profiling and reduction by plasma-activated water.: This research examines the distribution and reduction of Prochloraz residues and their metabolites like BTS44596 in citrus plants, contributing to the understanding of fungicide degradation products in environmental impact assessments (Lin et al., 2023).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Prochloraz Metabolite BTS44596


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library