Skip to Content
Merck
All Photos(1)

Key Documents

34490

Sigma-Aldrich

Dibutylamine

puriss., ≥99.0% (GC)

Synonym(s):

N-Butyl-1-butanamine, n-Dibutylamine, Di-n-butylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)2NH
CAS Number:
Molecular Weight:
129.24
Beilstein:
506001
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.46 (vs air)

Quality Level

vapor pressure

1.9 mmHg ( 20 °C)

grade

puriss.

Assay

≥99.0% (GC)

autoignition temp.

594 °F

expl. lim.

10 %

refractive index

n20/D 1.417 (lit.)
n20/D 1.417

bp

159 °C (lit.)

mp

−62 °C (lit.)

solubility

water: soluble 3.8 g/L at 20 °C

density

0.767 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCNCCCC

InChI

1S/C8H19N/c1-3-5-7-9-8-6-4-2/h9H,3-8H2,1-2H3

InChI key

JQVDAXLFBXTEQA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Dibutylamine was employed as organocatalyst during the synthesis of 2-amino-3-cyano-4H-chromen-4-ylphosphonates via Knoevenagel, Pinner and phospha-Michael reactions. Di-n-butylamine (Dibutylamine) may be used to investigate the performance of a dry sampler, with an impregnated denuder in series with a glass fibre filter for airborne isocyanates. It was used in the preparation of 1M dibutylammonium phosphate buffer.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

104.9 °F - closed cup

Flash Point(C)

40.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Reddi Mohan Naidu Kalla et al.
European journal of medicinal chemistry, 76, 61-66 (2014-02-27)
A series of 2-amino-3-cyano-4H-chromen-4-ylphosphonates have been synthesized by reacting substituted salicylaldehydes, malononitrile, and dialkylphosphites using a catalytic amount of dibutylamine as an organocatalyst employing Knoevenagel, Pinner, and phospha-Michael reactions simultaneously in ethanol. This protocol is an environmentally friendly procedure and
Daniel Gylestam et al.
The Annals of occupational hygiene, 58(1), 28-49 (2013-08-21)
The performance of a dry sampler, with an impregnated denuder in series with a glass fibre filter, using di-n-butylamine (DBA) for airborne isocyanates (200ml min(-1)) is investigated and compared with an impinger flask with a glass fibre filter in series
C M Stobba-Wiley et al.
Journal of AOAC International, 83(4), 837-846 (2000-09-20)
A method was developed and validated for determination and quantitation of tilmicosin residues in swine, cattle, and sheep edible tissues, as well as chicken fat, skin, and muscle over a concentration range of 0.025 microg/g-20 microg/g. For chicken kidney and
Baiyi Xue et al.
Journal of mass spectrometry : JMS, 46(7), 689-695 (2011-07-12)
Characterization of glycosaminoglycans poses a challenge for current analytical techniques, as they are highly acidic, polydisperse and heterogeneous compounds. The purpose of this study is the separation and analysis of a partially depolymerized heparin-like glycosaminoglycan by on-line ion-pairing reversed-phase high-performance
J T Fitzsimons et al.
Comparative biochemistry and physiology. A, Comparative physiology, 93(1), 285-290 (1989-01-01)
1. Ultrastructural changes in liver tissue of mice fed nitrosamine precursors, dibutylamine and nitrite, were observed. 2. The protective effect of soyabean in a diet containing nitrosamine precursors was demonstrated. 3. Liver tissue was examined to investigate the anticarcinogenicity of

Articles

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Knoevenagel Condensation is an organic reaction named after Emil Knoevenagel. It is a classic C-C bond formation reaction and a modification of the Aldol Condensation.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service