Skip to Content
Merck
All Photos(2)

Key Documents

34371

Supelco

Propetamphos

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20NO4PS
CAS Number:
Molecular Weight:
281.31
Beilstein:
1979853
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CCNP(=S)(OC)O\C(C)=C\C(=O)OC(C)C

InChI

1S/C10H20NO4PS/c1-6-11-16(17,13-5)15-9(4)7-10(12)14-8(2)3/h7-8H,6H2,1-5H3,(H,11,17)/b9-7+

InChI key

BZNDWPRGXNILMS-VQHVLOKHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Propetamphos is an organophosphate insecticide, which acts by inhibiting acetylcholinesterase (AChE) enzyme activity.

Application

Propetamphos may be used as an analytical reference standard for the determination of the analyte in human tissues, fruits and crops by various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A A Enayati et al.
Veterinary parasitology, 162(1-2), 135-141 (2009-03-17)
Hard tick infestation is an important problem in both traditional and industrial animal husbandry as well as in veterinary and public health. Several acaricides have been used to control tick infestation in Iran. Poor control has been attributed to acaricide
G C Coles et al.
Veterinary parasitology, 83(3-4), 327-330 (1999-07-28)
The response of sheep scab mites to pyrethroid insecticides and organophosphate compounds was studied in vitro with the objective of finding a simple test for detecting insecticide resistance in scab mites. Psoroptes cuniculi from rabbits or P. ovis from sheep
W A Virtue et al.
The Science of the total environment, 194-195, 207-217 (1997-02-24)
The Tweed River Purification Board's objective of reducing the numbers and significance of water pollution incidents by a proactive approach based on persuasion and education is described. This has consisted of prioritising potential pollutant sources which have then been investigated
S J Garfitt et al.
Biomarkers : biochemical indicators of exposure, response, and susceptibility to chemicals, 7(2), 113-122 (2002-07-10)
Propetamphos is a member of the vinyl phosphate group of insecticides and is mainly used for sheep dipping. There have been no published metabolic studies on the effect of propetamphos in man to date, although the present authors have published
'Sheep flu'.
CDR (London, England : Weekly), 1(42), 191-191 (1991-10-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service