Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C15H17ClN4
CAS Number:
Molecular Weight:
288.78
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
7138849
eCl@ss:
25140904
grade
analytical standard
Quality Segment
product line
PESTANAL®
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
application(s)
agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care
format
neat
SMILES string
CCCCC(Cn1cncn1)(C#N)c2ccc(Cl)cc2
InChI
1S/C15H17ClN4/c1-2-3-8-15(9-17,10-20-12-18-11-19-20)13-4-6-14(16)7-5-13/h4-7,11-12H,2-3,8,10H2,1H3
InChI key
HZJKXKUJVSEEFU-UHFFFAOYSA-N
General description
Myclobutanil is a broad spectrum systemic triazole fungicide, which can serve as potent inhibitor of ergosterol biosynthesis and hence, can be used to prevent any fungal attack on plants. It mainly shows efficacy against powdery mildew of vegetables and cereals.
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Legal Information
PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Myclobutanil
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Repr. 2
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
>212.0 °F
Flash Point (°C)
> 100 °C
PPE (personal protective equipment)
Eyeshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


