Skip to Content
Merck
All Photos(2)

Key Documents

34309

Supelco

Heptachlor exo-epoxide

PESTANAL®, analytical standard, isomer B

Synonym(s):

cis-Heptachlor epoxide, exo-1,4,5,6,7,8,8-Heptachloro-2,3-epoxy-4,7-methano-3a,4,7,7a-tetrahydroindane, HCE

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H5Cl7O
CAS Number:
Molecular Weight:
389.32
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Cl[C@H]1[C@H]2O[C@H]2[C@H]3[C@@H]1C4(Cl)C(Cl)=C(Cl)C3(Cl)C4(Cl)Cl

InChI

1S/C10H5Cl7O/c11-3-1-2(4-5(3)18-4)9(15)7(13)6(12)8(1,14)10(9,16)17/h1-5H/t1-,2+,3+,4-,5+,8?,9?/m0/s1

InChI key

ZXFXBSWRVIQKOD-TTWWGNBVSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chhatra Mani Sharma et al.
Environmental pollution (Barking, Essex : 1987), 157(8-9), 2452-2458 (2009-03-31)
Organochlorine pollutants in the major fish species (pike Esox lucius, perch Perca fluviatilis, and roach Rutilus rutilus) of Lake Arungen, Norway, were investigated after an extensive removal of large pike in 2004. The organochlorine pollutants detected in fish liver samples
Ulrike Luderer et al.
Environmental research, 121, 84-94 (2012-12-01)
For 15 months in 1981-1982, the commercial milk supply on the Hawaiian island of Oahu was contaminated with heptachlor epoxide, a metabolite of the insecticide heptachlor, resulting in gestational and/or lactational exposure to offspring of women who drank cow milk
M E Hansen et al.
Toxicology, 160(1-3), 139-153 (2001-03-14)
The effects of the organochlorine (OC) liver tumor promoter heptachlor epoxide (HE; 0, 0.1, 1, 10, and 50 microM) on several cellular tumor promoter-sensitive parameters were studied in mouse 1c1c7 hepatoma cells in an effort to identify the most sensitive
J P Hickey et al.
Archives of environmental contamination and toxicology, 50(1), 97-110 (2005-12-06)
Levels of chlorinated organic contaminants in predator fish have been monitored annually in each of the Great Lakes since the 1970s. This article updates earlier reports with data from 1991 to 1998 for lake trout (Salvelinus namaycush) and (Lake Erie
Frank H Pierik et al.
Environmental research, 105(3), 364-369 (2007-05-29)
Prenatal exposure to environmental endocrine disrupters has been postulated to cause adverse effects on male reproductive health. Exposure to organochlorine pesticides with anti-androgenic and estrogenic potency has been shown to interfere with the sex-hormone-dependent process of testicular descent in animal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service