Skip to Content
Merck
All Photos(2)

Key Documents

34306

Supelco

Quizalofop-ethyl

PESTANAL®, analytical standard

Synonym(s):

Ethyl 2-[4-(6-chloro-2-quinoxalinyloxy)phenoxy]propionate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17ClN2O4
CAS Number:
Molecular Weight:
372.80
Beilstein:
7145610
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCOC(=O)C(C)Oc1ccc(Oc2cnc3cc(Cl)ccc3n2)cc1

InChI

1S/C19H17ClN2O4/c1-3-24-19(23)12(2)25-14-5-7-15(8-6-14)26-18-11-21-17-10-13(20)4-9-16(17)22-18/h4-12H,3H2,1-2H3

InChI key

OSUHJPCHFDQAIT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quizalofop-ethyl is a phenoxy propionate herbicide, which is used to control both annual and perennial grass weeds in broadleaved crops.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of [14C] quizalofop-ethyl in soybean and cotton plants
Koeppe.KM, et al.
Journal of Agricultural and Food Chemistry, 38, 1085-1091 (1990)
Residual behavior of quizalofop ethyl on onion (Allium cepa L.)
Sahoo.K.S, et al.
Environmental Monitoring and Assessment, 185, 1711-1718 (2013)
S K Sahoo et al.
Environmental monitoring and assessment, 185(2), 1711-1718 (2012-05-11)
Quizalofop ethyl, a phenoxy propionate herbicide, is used for postemergence control of annual and perennial grass weeds in broad-leaved crops in India. The experiments were designed to study the dissipation kinetics of quizalofop ethyl on onion for two seasons. A
Ioannis S Elefsiniotis et al.
Environmental health perspectives, 115(10), 1479-1481 (2007-10-17)
Quizalofop-p-ethyl is an often applied, slightly toxic herbicide for which no severe toxicity has been reported in humans. We present the case of a farmer exposed to quizalofop-p-ethyl who presented with obstructive cholestasis. A complete workup disclosed no other cause
Xiaoxiang Zhang et al.
Journal of hazardous materials, 186(1), 876-882 (2010-12-17)
Transformation products usually differ in environmental and toxicological properties compared to the parent contaminants, thus causing potential and unknown environmental risks. To elucidate differential chiral recognition of the aryloxypropanoate herbicide quizalofop ethyl (QE) and its primary product (quizalofop acid, QA)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service