Skip to Content
Merck

34036

Aflatoxin Mix 4 solution

analytical standard

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
85151701
EC Number:
200-835-2
NACRES:
NA.24
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

shelf life

limited shelf life, expiry date on the label

concentration

0.5 μg/mL B2 and G2 in acetonitrile, 2 μg/mL B1 and G1 in acetonitrile

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

multi-component solution

storage temp.

−20°C

General description

Aflatoxins are a group of hypertoxic secondary metabolites produced by fungi of the genus Aspergillus, mostly A. flavus, A. Parasiticus, and A. nomius. Of the 20 aflatoxins identified so far, B1, B2, G1, and G2 are the four naturally occurring aflatoxins.

Application

It is intended to be used for the calibration of analytical instruments, validation of analytical methods, determination of detection limits and linearity studies, and recovery experiments.

This aflatoxin mix 4 solution was used as follows:
  • Ultra-high-performance liquid chromatography-fluorescence detection (UHPLC-FLD) based quantification of aflatoxins— B1, B2, G1, and G2, in the coffee bean and powder samples
  • Estimation of aflatoxins (B1, B2, G1, and G2) in samples of maize, rice, sorghum meal, and peanuts by ultra high-performance liquid chromatography coupled with a fluorescence detector
  • Determination of naturally occurring aflatoxins in 65 peanut-enriched flour samples by high-performance liquid chromatography (HPLC) combined with fluorescence detection

Other Notes

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Disclaimer

Supply conditions do not apply to the regions and states of Brazil: North, Northeast, Mato Grosso do Sul, Mato Grosso, and Rio Grande do Sul.


Still not finding the right product?

Explore all of our products under Aflatoxin Mix 4 solution


pictograms

FlameExclamation mark

signalword

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 2

Storage Class

3 - Flammable liquids

wgk

WGK 2

flash_point_f

35.6 °F - closed cup

flash_point_c

2 °C - closed cup

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library