Skip to Content
Merck
All Photos(1)

Key Documents

34002

Supelco

(S)-(−)-Sulpiride

VETRANAL®, analytical standard

Synonym(s):

(S)-5-Aminosulfonyl-N-[(1-ethyl-2-pyrrolidinyl)methyl]-2-methoxybenzamide, Levosulpiride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H23N3O4S
CAS Number:
Molecular Weight:
341.43
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

183-186 °C (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CCN1CCC[C@H]1CNC(=O)c2cc(ccc2OC)S(N)(=O)=O

InChI

1S/C15H23N3O4S/c1-3-18-8-4-5-11(18)10-17-15(19)13-9-12(23(16,20)21)6-7-14(13)22-2/h6-7,9,11H,3-5,8,10H2,1-2H3,(H,17,19)(H2,16,20,21)/t11-/m0/s1

InChI key

BGRJTUBHPOOWDU-NSHDSACASA-N

Looking for similar products? Visit Product Comparison Guide

General description

(S)-(−)-Sulpiride is a levorotatory enantiomer of sulpiride, which shows greater central antidopaminergic activity, antiemetic and antidyspeptic effects. It is also used as a therapeutic drug for the treatment of depression and somatoform disorders.

Application

(S)-(−)-Sulpiride may be used as an analytical reference standard for the determination of the drug, in biological samples using hydrophilic interaction liquid chromatography−tandem mass spectrometry (HILIC−MS/MS) and reversed-phase high-performance liquid chromatography with fluorescence detection (HPLC-FD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

D2 dopamine receptor antagonist; antipsychotic.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Levosulpiride: a review of its clinical use in psychiatry.
Mucci A, et al.
Pharmacological Research, 31(2), 95-101 (1995)
Improvement and validation of a liquid chromatographic method for the determination of levosulpiride in human serum and urine.
Cho Y-H and Lee B-Y
Journal of Chromatography. B, Biomedical Sciences and Applications, 976(2), 243-251 (2003)
Type 1 (insulin-dependent) diabetes mellitus and gastric emptying: role of blood glucose concentration and dopamine D2 receptors.
P Melga et al.
Diabetes, nutrition & metabolism, 17(1), 62-65 (2004-05-28)
Hyuk Lee et al.
Journal of gastroenterology and hepatology, 22(11), 1741-1747 (2007-10-05)
This prospective study was conducted to characterize the clinical features of adult rumination syndrome, in the absence of psychiatric illness, by applying newly modified clinical criteria, and to elucidate factors influencing outcomes after treatment. Twenty-one adult patients diagnosed with rumination
Raúl León-Barúa et al.
Revista de gastroenterologia del Peru : organo oficial de la Sociedad de Gastroenterologia del Peru, 28(3), 267-269 (2008-10-30)
We expose, this time, our ideas and findings on the etiopathogenesis, diagnosis and treatment of the dyspeptic syndrome.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service