Skip to Content
Merck
All Photos(1)

Key Documents

33976

Supelco

Ethiprole

PESTANAL®, analytical standard

Synonym(s):

5-Amino-1-(2,6-dichloro-α,α,α-trifluoro-p-tolyl)-4-(ethylsulfinyl)pyrazole-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H9Cl2F3N4OS
CAS Number:
Molecular Weight:
397.20
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

CCS(=O)c1c(N)n(nc1C#N)-c2c(Cl)cc(cc2Cl)C(F)(F)F

InChI

1S/C13H9Cl2F3N4OS/c1-2-24(23)11-9(5-19)21-22(12(11)20)10-7(14)3-6(4-8(10)15)13(16,17)18/h3-4H,2,20H2,1H3

InChI key

FNELVJVBIYMIMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Ethiprole, is a pyrazole insecticide, which can be used to control a broad spectrum of insects(1), like those with chewing and sucking mouthparts that tend to damage the plants. It can be mainly used for rice and cotton cultivation.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2 Oral

Target Organs

Liver,Thyroid

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Kaiying Chen et al.
Water research, 161, 531-539 (2019-06-24)
Ethiprole and its degradation products were identified and quantified using UHPLC-Q-TOF/MS. Indoor culture experiments and UHPLC-Q-TOF/MS are used to study the hydrolysis, photolysis, and soil degradation behavior of ethiprole in water and soil. The differences in ethiprole degradation products across
Efficacy of ethiprole applied alone and in combination with conventional insecticides for protection of stored wheat and stored corn
Arthur HF
Journal of Economic Entomology, 95, 1314-1318 (2002)
Fluorine and the Environment: Agrochemicals, Archaeology, Green Chemistry and Water (2006)
Yueyue Liu et al.
Environmental geochemistry and health, 43(1), 461-473 (2020-10-08)
Ethiprole has been widely used in agriculture, but there have been few studies on the adverse effects of ethiprole on nontarget organisms. This study focused on the mechanism of the sublethal effects of ethiprole on the development, antioxidation mechanisms, detoxification
The Toxicology and Biochemistry of Insecticides, Second Edition (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service