Skip to Content
Merck

33882

SCA-13C-15N2 hydrochloride

VETRANAL®, analytical standard

Synonym(s):

Semicarbazide-13C-15N2 hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
13CH5N15N2O · HCl
CAS Number:
Molecular Weight:
114.51
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

clinical testing

format

neat

mass shift

M+3

SMILES string

Cl.N[13C](=O)[15NH][15NH2]

InChI

1S/CH5N3O.ClH/c2-1(5)4-3;/h3H2,(H3,2,4,5);1H/i1+1,3+1,4+1;

InChI key

XHQYBDSXTDXSHY-XRNBZBNOSA-N

General description

SCA-13C-15N2 hydrochloride is an isotopically labeled metabolite of nitrofuran, an antimicrobial drug used in veterinary medicine.

Application

SCA-13C-15N2 hydrochloride has been used as an internal standard for the determination of semicarbazide in aquaculture products and honey samples using various chromatography techniques.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Oral

target_organs

Bone

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library