Skip to Content
Merck
All Photos(1)

Key Documents

33864

Supelco

Tilmicosin

VETRANAL®, analytical standard, mix of isomers

Synonym(s):

20-Deoxo-20-(3,5-dimethyl-1-piperidinyl)desmycosin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C46H80N2O13
Molecular Weight:
869.13
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CC1OC(OCC(C(CC)O2)/C=C(/C=C/C(C(C)CC(CCN3CC(C)CC(C)C3)C(OC4OC(C)C(O)C(N(C)C)C4O)C(C)C(O)CC2=O)=O)C)C(OC)C(OC)C1O

InChI

1S/C46H80N2O13/c1-13-36-33(24-57-46-44(56-12)43(55-11)40(53)31(8)59-46)19-25(2)14-15-34(49)28(5)20-32(16-17-48-22-26(3)18-27(4)23-48)42(29(6)35(50)21-37(51)60-36)61-45-41(54)38(47(9)10)39(52)30(7)58-45/h14-15,19,26-33,35-36,38-46,50,52-54H,13,16-18,20-24H2,1-12H3/b15-14+,25-19+/t26-,27+,28-,29+,30-,31-,32+,33-,35-,36-,38+,39-,40-,41-,42-,43-,44-,45+,46-/m1/s1

InChI key

JTSDBFGMPLKDCD-XVFHVFLVSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Tilmicosin is a semisynthetic macrolide antibiotic, with broad spectrum activity towards Mycoplasma spp., Pasteurella spp., and various gram-positive organisms in animals.

Application

Commission Regulation (EU) No 37/2010 of 22 December 2009 on pharmacologically active substances and their classification regarding maximum residue limits in foodstuffs of animal origin
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Tilmicosin may be used as a reference standard for the determination of the analyte in chicken, swine, cattle, sheep tissues and milk samples using liquid chromatography with ultraviolet detection (LC-UV).

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - STOT RE 2

Target Organs

Cardio-vascular system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niels Møller Andersen et al.
Antimicrobial agents and chemotherapy, 56(11), 6033-6036 (2012-08-29)
Tildipirosin is a 16-membered-ring macrolide developed to treat bacterial pathogens, including Mannheimia haemolytica and Pasteurella multocida, that cause respiratory tract infections in cattle and swine. Here we evaluated the efficacy of tildipirosin at inhibiting protein synthesis on the ribosome (50%
Jacob Poehlsgaard et al.
ACS chemical biology, 7(8), 1351-1355 (2012-05-09)
The veterinary antibiotic tildipirosin (20,23-dipiperidinyl-mycaminosyl-tylonolide, Zuprevo) was developed recently to treat bovine and swine respiratory tract infections caused by bacterial pathogens such as Pasteurella multocida. Tildipirosin is a derivative of the naturally occurring compound tylosin. Here, we define drug-target interactions
Yingyu Liu et al.
Microbial drug resistance (Larchmont, N.Y.), 18(6), 604-615 (2012-09-01)
The Haemophilus parasuis respiratory tract pathogen poses a severe threat to the swine industry despite available antimicrobial therapies. To gain a more detailed understanding of the molecular mechanisms underlying H. parasuis response to tilmicosin treatment, microarray technology was applied to
P Bosi et al.
Journal of animal science, 89(12), 4043-4053 (2011-07-05)
The aim of this study was to determine the effects of 3 antibiotics used for pulmonary pathologies added in the feed of weaned pigs on growth performance, commensal microbiota, and immune response. At weaning, a total of 72 pigs were
Xinxin Ci et al.
Immunopharmacology and immunotoxicology, 33(4), 626-632 (2011-03-25)
Tilmicosin, a semi-synthetic tylosin-derived macrolide antibiotic commonly used by veterinarians, has been shown to possess anti-inflammatory activity. However, possible use in asthma treatment has not yet been studied. In this study, we investigated the anti-inflammatory properties of tilmicosin using a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service