Skip to Content
Merck

Skip To

33656

Semicarbazide hydrochloride

VETRANAL®, analytical standard

Synonym(s):

N-Aminourea hydrochloride, Hydrazine carboxamide hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
NH2CONHNH2 · HCl
CAS Number:
Molecular Weight:
111.53
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
209-247-0
Beilstein/REAXYS Number:
3593642
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

mp

175-177 °C (dec.) (lit.)

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

Cl.NNC(N)=O

InChI

1S/CH5N3O.ClH/c2-1(5)4-3;/h3H2,(H3,2,4,5);1H

InChI key

XHQYBDSXTDXSHY-UHFFFAOYSA-N

Application

Semicarbazide hydrochloride may be used in the synthesis of 4-nitrobenzaldehyde semicarbazone via reaction with 4-nitrobenzaldehyde.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Semicarbazide hydrochloride may be used as a stabilizing agent for the determination of norepinephrine in plasma samples using stable-isotope dilution gas chromatography-tandem mass spectrometry with negative ion chemical ionization.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
S2201D0638PHR3184
description

VETRANAL®, analytical standard

description

≥99%

description

analytical standard, ≥99%

description

pharmaceutical secondary standard, certified reference material

grade

analytical standard

grade

-

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

format

neat

format

-

format

neat

format

-

product line

VETRANAL®

product line

-

product line

-

product line

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

Quality Level

100

Quality Level

200

Quality Level

200

Quality Level

300

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-


Still not finding the right product?

Explore all of our products under Semicarbazide hydrochloride


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - Repr. 2 - Skin Corr. 1B - STOT RE 2 Oral

target_organs

Bone

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P2 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters