Skip to Content
Merck
All Photos(1)

Key Documents

33576

Supelco

Microcystin-YR solution

10 μg/mL in methanol, analytical standard

Synonym(s):

Algae bloom toxin, Biotoxin, Cyanobacterial toxin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H72N10O13
CAS Number:
Molecular Weight:
1045.19
EC Number:
UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

shelf life

limited shelf life, expiry date on the label

concentration

10 μg/mL in methanol

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

single component solution

storage temp.

−20°C

SMILES string

CO[C@@H](Cc1ccccc1)[C@@H](C)\C=C(C)\C=C\[C@@H]2NC(=O)[C@H](CCCNC(N)=N)NC(=O)[C@@H](C)[C@@H](NC(=O)[C@H](Cc3ccc(O)cc3)NC(=O)[C@@H](C)NC(=O)C(=C)N(C)C(=O)CC[C@@H](NC(=O)[C@H]2C)C(O)=O)C(O)=O

InChI

1S/C52H72N10O13/c1-28(25-29(2)41(75-8)27-34-13-10-9-11-14-34)16-21-37-30(3)44(65)59-39(50(71)72)22-23-42(64)62(7)33(6)47(68)56-32(5)46(67)60-40(26-35-17-19-36(63)20-18-35)49(70)61-43(51(73)74)31(4)45(66)58-38(48(69)57-37)15-12-24-55-52(53)54/h9-11,13-14,16-21,25,29-32,37-41,43,63H,6,12,15,22-24,26-27H2,1-5,7-8H3,(H,56,68)(H,57,69)(H,58,66)(H,59,65)(H,60,67)(H,61,70)(H,71,72)(H,73,74)(H4,53,54,55)/b21-16+,28-25+/t29-,30-,31-,32+,37-,38-,39+,40-,41-,43+/m0/s1

InChI key

OWHASZQTEFAUJC-GJRPNUFSSA-N

General description

Microcystin-YR is a microcystin that is generally produced by cyanobacteria.

Application

Microcystin-YR may be used as an analytical reference standard for the quantification of the analyte in water samples using high-performance liquid chromatography with diode array detection (HPLC-DAD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

51.8 °F

Flash Point(C)

11 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nidhi Gupta et al.
Toxicology, 188(2-3), 285-296 (2003-05-28)
The cyclic peptide toxins microcystins and nodularins are the most common and abundant cyanotoxins present in diverse water systems. They have been the cause of human and animal health hazards and even death. Over 60 microcystin variants have been reported
Dawen Zhang et al.
The Science of the total environment, 407(7), 2191-2199 (2009-02-03)
In this paper, accumulation and distribution of microcystins (MCs) was examined monthly in six species of fish with different trophic levels in Meiliang Bay, Lake Taihu, China, from June to November 2005. Microcystins were analyzed by liquid chromatography electrospray ionization
Douglas O Mountfort et al.
Toxicon : official journal of the International Society on Toxinology, 45(2), 199-206 (2005-01-01)
Depending on the class of microcystin the protein phosphatase inhibition assay shows different sensitivities to different classes of toxin. We have determined that the IC50 values obtained from dose-response curves for the inhibition of the enzyme by micro-cystin LR, nodularin
Joanna Mankiewicz et al.
Environmental toxicology, 20(5), 499-506 (2005-09-15)
The lakes of northern Poland are among the recreational sites most valued by Polish and German holiday makers. Given the socioeconomic importance of these lakes, water quality should be maintained at high levels for such intensive recreational purposes. In 2002
M Douma et al.
Environmental monitoring and assessment, 160(1-4), 439-450 (2009-01-09)
Mansour Eddahbi (MED) (30 degrees 55'N, 6 degrees 53'W) and Almassira (ALM) (31 degrees 95'N, 6 degrees 72'W) are two Moroccan lake reservoirs located at an arid and semi-arid hydrographic basin, respectively. Both are used for irrigation, recreational activities and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service