Skip to Content
Merck
All Photos(2)

Key Documents

33496

Supelco

Flucythrinate

PESTANAL®, analytical standard, mixture of stereo isomers

Synonym(s):

α-Cyano-3-phenoxybenzyl 4-difluoromethoxy-α-isopropylphenylacetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C26H23F2NO4
CAS Number:
Molecular Weight:
451.46
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

CC(C)C(C(=O)OC(C#N)c1cccc(Oc2ccccc2)c1)c3ccc(OC(F)F)cc3

InChI

1S/C26H23F2NO4/c1-17(2)24(18-11-13-21(14-12-18)32-26(27)28)25(30)33-23(16-29)19-7-6-10-22(15-19)31-20-8-4-3-5-9-20/h3-15,17,23-24,26H,1-2H3

InChI key

GBIHOLCMZGAKNG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flucythrinate belongs to the class of synthetic pyrethroids, primarily used to control Heliothis spp. in cotton and other insect pests in agricultural crops.

Application

Flucythrinate may be used as a reference standard for the determination of the analyte in milk samples using gas chromatography with electron capture detection (GC-ECD).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

113.0 °F

Flash Point(C)

45 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evaluation of flucythrinate- and fenvalerate-impregnated ear tags and permethrin ear tapes for fly (Diptera: Muscidae) control on beef and dairy cattle in northwest Florida.
J A Hogsette et al.
Journal of economic entomology, 79(1), 152-157 (1986-02-01)
Pesticide Profiles: Toxicity, Environmental Impact, and Fate (1997)
L K Cutkomp et al.
Journal of the American Mosquito Control Association, 2(3), 347-349 (1986-09-01)
The influence of temperature on the toxicity of the pyrethroids cypermethrin, permethrin, fenvalerate, d-phenothrin, flucythrinate and bioallethrin to 3rd instar Aedes aegypti larvae was determined. Based on LC50 levels, the toxicities of all pyrethroids were in the range of 1.33-
Comparison of dislodgable and total residues of three pyrethroids applied to cotton in Arizona.
B J Estesen et al.
Bulletin of environmental contamination and toxicology, 44(2), 240-245 (1990-02-01)
S A Flannigan et al.
British journal of industrial medicine, 42(11), 773-776 (1985-11-01)
Synthetic pyrethroids are neurotoxic insecticides with a low mammalian toxicity. Prior investigations have found these agents to be neither cutaneous irritants nor sensitisers. Clinically demonstrable inflammation, as judged by erythema, oedema, or vesiculation, has not been apparent. Nevertheless, paraesthesia does

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service