Skip to Content
Merck
All Photos(2)

Documents

32979

Supelco

Acetamiprid-N-desmethyl

PESTANAL®, analytical standard

Synonym(s):

N-(6-Chloro-3-pyridylmethyl)-N′-cyano-acetamidine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9ClN4
CAS Number:
Molecular Weight:
208.65
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

C\C(NCc1ccc(Cl)nc1)=N/C#N

InChI

1S/C9H9ClN4/c1-7(14-6-11)12-4-8-2-3-9(10)13-5-8/h2-3,5H,4H2,1H3,(H,12,14)

InChI key

AYEAUPRZTZWBBF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetamiprid-N-desmethyl is a metabolite of the neonicotinoid insecticide, acetamiprid.

Application

Acetamiprid-N-desmethyl may be used as a reference standard in the determination of acetamiprid-N-desmethyl in urine samples using ultra high-performance liquid chromatography coupled with orbitrap high resolution mass spectrometry (UHPLC-HRMS).
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Acetamipride-Metabolite IM-2-1

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Richard C Kolanczyk et al.
Xenobiotica; the fate of foreign compounds in biological systems, 50(7), 805-814 (2019-11-15)
Providing an alternative to pyrethroids, organophosphates, and carbamates, the neonicotinoids are now the most widely used insecticides in the world. They are water soluble and relatively stable in soil and water which allows for run-off through surface waters and thus
Relationship between urinary N-desmethyl-acetamiprid and typical symptoms including neurological findings: a prevalence case-control study
Marfo TJ, et al.
PLoS ONE, 10(11), e0142172-e0142172 (2015)
Takamitsu Otake et al.
Journal of AOAC International, 103(6), 1469-1477 (2020-11-29)
Neonicotinoid insecticides (neonicotinoids) are widely used, however, they can negatively affect human health. Thus, neonicotinoids and their metabolites in human urine are analyzed globally to assess exposure levels and health risks. To properly assess the exposure levels and health risks
Marina López-García et al.
Journal of pharmaceutical and biomedical analysis, 146, 378-386 (2017-09-18)
A semiautomated method based on ultra-high performance liquid chromatography (UHPLC) coupled to Orbitrap high resolution mass spectrometry has been developed for the determination of neonicotinoids (imidacloprid, acetamiprid, clothianidin, dinotefuran, nitenpyram, thiacloprid and thiamethoxam) and the metabolite acetamiprid-n-desmethyl in urine samples.
Mohammad Faraji et al.
Food chemistry, 240, 634-641 (2017-09-28)
A QuEChERS based methodology was developed for the simultaneous identification and quantification of acetamiprid, imidacloprid, and spirotetramat and their relevant metabolites in pistachio by liquid chromatography-tandem mass spectrometry for the first time. First, sample extraction was done with MeCN:citrate buffer:NaHCO

Protocols

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

Learn more about Neonicotinoids - active substances used in plant protection products to control harmful insects.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service