Skip to Content
Merck
All Photos(1)

Key Documents

32801

Supelco

Flubendiamide

PESTANAL®, analytical standard

Synonym(s):

N2-[1,1-Dimethyl-2-(methylsulfonyl)ethyl]-3-iodo-N1-{2-methyl-4-[1,2,2,2-tetrafluoro-1-(trifluoromethyl)ethyl]phenyl}-1,2-benzenedicarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H22F7IN2O4S
CAS Number:
Molecular Weight:
682.39
Beilstein:
11326832
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

Cc1cc(ccc1NC(=O)c2cccc(I)c2C(=O)NC(C)(C)CS(C)(=O)=O)C(F)(C(F)(F)F)C(F)(F)F

InChI

1S/C23H22F7IN2O4S/c1-12-10-13(21(24,22(25,26)27)23(28,29)30)8-9-16(12)32-18(34)14-6-5-7-15(31)17(14)19(35)33-20(2,3)11-38(4,36)37/h5-10H,11H2,1-4H3,(H,32,34)(H,33,35)

InChI key

ZGNITFSDLCMLGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Flubendiamide is a novel insecticide that is grouped under the family of phthalic acid diamides. It is largely used against lepidopteron pests in various annual and perennial crops.

Application

Flubendiamide may be used as an analytical reference standard for the determination of the analyte in vegetables and fruits by high performance liquid chromatography coupled to ultraviolet detector (HPLC-UV) and HPLC combined with tandem mass spectrometry (MS/MS), respectively.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ana M Ares et al.
Food chemistry, 232, 169-176 (2017-05-12)
In this study, a new method has been developed to determine flubendiamide in honey using liquid chromatography coupled to a selective mass spectrometry detector (quadrupole-time-of-flight). An efficient sample treatment involving a solid phase extraction with a C
Shun-Fan Wu et al.
Pesticide biochemistry and physiology, 146, 1-6 (2018-04-09)
The susceptibilities of three field populations of pink stem borer (PSB), Sesamia inferens (walker) to diamide insecticides, chlorantraniliprole and flubendiamide, were evaluated in this study. The results showed that these PSB field populations were still sensitive to the two diamide
A fast method for determination of flubendiamide in vegetables by liquid chromatography.
Mukherjee I, et al.
Pesticide Research Journal, 24(2), 159-162 (2012)
Simple and efficient method for the estimation of residues of flubendiamide and its metabolite desiodo flubendiamide.
Singh B, et al.
Journal of Agricultural and Food Chemistry, 56(7), 2299-2302 (2008)
Shaon Kumar Das et al.
Bulletin of environmental contamination and toxicology, 105(2), 261-269 (2020-07-16)
Persistence and sorption behaviour of flubendiamide in two different Indian soils as affected by maize stalk biochar was studied. The persistence was more in West Bengal soil (178.6 days) than Sikkim soil (165.3 days) at 10 µg g-1 fortification level. Biochar

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service