Skip to Content
Merck
All Photos(2)

Key Documents

32593

Supelco

Cambendazole

VETRANAL®, analytical standard

Synonym(s):

Isopropyl 2-(4-thiazolyl)-5-benzimidazolecarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14N4O2S
CAS Number:
Molecular Weight:
302.35
Beilstein:
563292
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

product line

VETRANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

SMILES string

CC(C)OC(=O)Nc1ccc2[nH]c(nc2c1)-c3cscn3

InChI

1S/C14H14N4O2S/c1-8(2)20-14(19)16-9-3-4-10-11(5-9)18-13(17-10)12-6-21-7-15-12/h3-8H,1-2H3,(H,16,19)(H,17,18)

InChI key

QZWHWHNCPFEXLL-UHFFFAOYSA-N

General description

Cambendazole is classified under the family of benzimidazole anthelmintic compounds.[1]

Application

Cambendazole may be used as an analytical reference standard for the determination of the analyte in:
  • Bovine muscle samples by ultra-high performance liquid chromatography (UHPLC) coupled to tandem mass spectrometry (MS/MS) operating in ESI mode.[2]
  • Bovine milk samples by LC-ESI-MS.[3]

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Morphological changes in Capillaria obsignata (Nematoda: Trichuridae) eggs after treatment with cambendazole.
P Dorny et al.
Parasitology research, 74(2), 196-197 (1987-01-01)
[Trial of the modification of immunopathological changes in the muscular phase of trichinosis I. The effect of cambendazole].
K Karmańska et al.
Wiadomosci parazytologiczne, 33(4-5), 517-524 (1987-01-01)
[Evaluation of the therapeutic activity of albendazole in rats experimentally infected with Strongyloides venezuelensis].
V Amato Neto et al.
Revista do Instituto de Medicina Tropical de Sao Paulo, 28(3), 181-184 (1986-05-01)
R N Beech et al.
Genetics, 138(1), 103-110 (1994-09-01)
Benzimidazole anthelmintics are the most common chemotherapeutic agents used to remove intestinal helminths from farm animals. The development of drug resistance within helminth populations is wide-spread and can render these drugs essentially useless. The mechanism of benzimidazole resistance appears to
Determination of benzimidazoles and levamisole residues in milk by liquid chromatography-mass spectrometry: screening method development and validation.
Jedziniak P, et al.
Journal of Chromatography A, 1216(46), 8165-8172 (2009)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service