Skip to Content
Merck

32504

Bifenazate

PESTANAL®, analytical standard

Synonym(s):

N′-(4-Methoxybiphenyl-3-yl)hydrazinecarboxylic acid isopropyl ester

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H20N2O3
CAS Number:
Molecular Weight:
300.35
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
442-820-5
Beilstein/REAXYS Number:
9632665
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1ccc(cc1NNC(=O)OC(C)C)-c2ccccc2

InChI

1S/C17H20N2O3/c1-12(2)22-17(20)19-18-15-11-14(9-10-16(15)21-3)13-7-5-4-6-8-13/h4-12,18H,1-3H3,(H,19,20)

InChI key

VHLKTXFWDRXILV-UHFFFAOYSA-N

General description

Bifenazate is a selective carbazate acaricide primarily used for the control of phytophagous mites infesting agricultural and ornamental crops.

Application

Bifenazate may be used as a reference standard for the determination of bifenazate from edible oils (olive, soya and sunflower) by liquid chromatography-tandem mass spectrometry employing new generation materials for dispersive solid phase extraction clean-up.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany


Still not finding the right product?

Explore all of our products under Bifenazate


signalword

Warning

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT RE 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library