Skip to Content
Merck

32502

8-Quinolinol

puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99% (perchloric acid titration)

Synonym(s):

8-Hydroxyquinoline, 8-Oxychinolin, Oxine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H7NO
CAS Number:
Molecular Weight:
145.16
UNSPSC Code:
12352100
NACRES:
NA.21
PubChem Substance ID:
EC Number:
205-711-1
Beilstein/REAXYS Number:
114512
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

agency

reag. Ph. Eur.

grade

ACS reagent, puriss. p.a.

assay

≥99% (perchloric acid titration)

form

solid

technique(s)

titration: suitable

impurities

≤0.05% insoluble in ethanol

ign. residue

≤0.05% (as SO4)

mp

72.5-74.0 °C, 72.5-75 °C

anion traces

chloride (Cl-): ≤10 mg/kg, sulfate (SO42-): ≤100 mg/kg

suitability

passes test for suitability for Mg decision

antibiotic activity spectrum

fungi

mode of action

DNA synthesis | interferes, enzyme | inhibits

SMILES string

Oc1cccc2cccnc12

InChI

1S/C9H7NO/c11-8-5-1-3-7-4-2-6-10-9(7)8/h1-6,11H

InChI key

MCJGNVYPOGVAJF-UHFFFAOYSA-N

General description

Chemical structure: quinolone
8-Quinolinol (QOH) forms complexes with heavy metals [Pt(II), Pb(II), Bi(III), Ir(III)]. These complexes show long-lived phosphorescence and excited state absorption in solutions. It has been reported to form metal chelate complexes. Immobilized 8-quinolinol has been widely used in the preconcentration step during the extraction of metals.


Still not finding the right product?

Explore all of our products under 8-Quinolinol


signalword

Danger

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 1B - Skin Sens. 1

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library