Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C26H31Cl2N5O3
CAS Number:
Molecular Weight:
532.46
MDL number:
UNSPSC Code:
41116107
EC Number:
267-751-6
NACRES:
NA.24
eCl@ss:
25140904
grade
analytical standard
Quality Segment
product line
VETRANAL®
technique(s)
HPLC: suitable, chromatography: suitable, gas chromatography (GC): suitable
application(s)
forensics and toxicology
veterinary
format
neat
SMILES string
Clc1c(ccc(c1)Cl)[C@@]3(O[C@H](CO3)COc4ccc(cc4)N5CCN(CC5)C(C)C)C[n]2ncnc2
InChI
1S/C26H31Cl2N5O3/c1-19(2)31-9-11-32(12-10-31)21-4-6-22(7-5-21)34-14-23-15-35-26(36-23,16-33-18-29-17-30-33)24-8-3-20(27)13-25(24)28/h3-8,13,17-19,23H,9-12,14-16H2,1-2H3/t23-,26-/m0/s1
InChI key
BLSQLHNBWJLIBQ-OZXSUGGESA-N
Application
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Legal Information
VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany
Still not finding the right product?
Explore all of our products under Terconazole
Signal Word
Warning
Hazard Codes
Precautionary Statements
Hazard Classifications
Aquatic Chronic 4 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class
11 - Combustible Solids
WGK
WGK 3
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
