Skip to Content
Merck
All Photos(2)

Documents

32333

Supelco

Fipronil sulfone

PESTANAL®, analytical standard

Synonym(s):

5-Amino-1-[2,6-dichloro-4-(trifluoromethyl)phenyl]-4-[(trifluoromethyl)sulfonyl]-1H-pyrazole-3-carbonitrile

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H4Cl2F6N4O2S
CAS Number:
Molecular Weight:
453.15
Beilstein:
8729974
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

NC1=C(S(=O)(C(F)(F)F)=O)C(C#N)=NN1C2=C(Cl)C=C(C(F)(F)F)C=C2Cl

InChI

1S/C12H4Cl2F6N4O2S/c13-5-1-4(11(15,16)17)2-6(14)8(5)24-10(22)9(7(3-21)23-24)27(25,26)12(18,19)20/h1-2H,22H2

InChI key

LGHZJDKSVUTELU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Find more information here fipronil

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Romero et al.
Toxicology letters, 252, 42-49 (2016-04-14)
Fipronil is a broad spectrum insecticide from the phenyl pyrazole family, which targets GABA receptor. Limited information is available about the metabolite fipronil sulfone cytotoxic actions. This study examined in vitro neurotoxicity of fipronil and fipronil sulfone and evaluated Trolox
Hiroshi Jinguji et al.
PloS one, 13(7), e0200299-e0200299 (2018-07-12)
Dragonflies, Sympetrum spp., are indispensable to agriculture and are a central element of culture in Japan. However, S. frequens populations in rice paddy fields have declined in recent decades. Dragonfly larvae are predatory aquatic insects that feed on other organisms
Natalia Vasylieva et al.
Environment international, 103, 91-98 (2017-03-28)
Occupational medical surveillance is highly desirable in manufacturing facilities where exposure to chemicals is significant. The insecticide fipronil is generally considered safe for humans but with increasing use, exposure to fipronil is of concern. Identification of urinary metabolites of fipronil
Anudurga Gajendiran et al.
3 Biotech, 7(3), 212-212 (2017-07-02)
Fipronil is a phenylpryazole insecticide which is extensively used for the protection of agricultural yields. However, this insecticide poses various threats to  the environment. Therefore it is essential to develop an effective method to degrade or eliminate this pollutant from the 
Jayanthi Abraham et al.
Pesticide biochemistry and physiology, 155, 90-100 (2019-03-13)
Fipronil is a most widely used insecticide. It has been found to be highly toxic to insects with alarming decline in honey bees' population. It has shown to cause severe contamination of soil and water. The present study was undertaken

Protocols

A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone.

A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone.

A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone.

A short abstract on a method using QuEChERS and GC/MS/MS for determination of fipronil & fipronil sulfone.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service