Skip to Content
Merck
All Photos(2)

Key Documents

32236

Supelco

Firocoxib

VETRANAL®, analytical standard

Synonym(s):

3-Cyclopropylmethoxy-4-(4-methanesulfonylphenyl)-5,5-dimethyl-5H-furan-2-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H20O5S
CAS Number:
Molecular Weight:
336.40
Beilstein:
8335837
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

O=S(C1=CC=C(C2=C(OCC3CC3)C(OC2(C)C)=O)C=C1)(C)=O

InChI

1S/C17H20O5S/c1-17(2)14(12-6-8-13(9-7-12)23(3,19)20)15(16(18)22-17)21-10-11-4-5-11/h6-9,11H,4-5,10H2,1-3H3

InChI key

FULAPETWGIGNMT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

William G Ryan et al.
Veterinary therapeutics : research in applied veterinary medicine, 8(4), 263-271 (2008-01-10)
Veterinarians currently have a choice of six registered NSAIDs for use in the management of canine osteoarthritis. There is a need for data to clarify whether there is an increased risk of side effects occurring in the event of a
W Back et al.
Equine veterinary journal, 41(3), 309-312 (2009-05-28)
Lameness is a highly prevalent condition in horses and the principal cause of removal from athletic activity. In clinical studies to evaluate nonsteroidal anti-inflammatory drug therapies, force plates are commonly used to assess improvement of lameness objectively. To use a
Laura T Letendre et al.
American journal of veterinary research, 69(11), 1399-1405 (2008-11-05)
To determine pharmacokinetic parameters and variables, firocoxib concentrations in urine and plasma, urine-to-plasma ratios, and the urine depletion profile of firocoxib and to evaluate whether the pharmacokinetic behavior of firocoxib was governed by linear processes after multiple doses of firocoxib
A Autefage et al.
The Veterinary record, 168(23), 617-617 (2011-06-07)
Thirty-nine client-owned dogs with osteoarthritis were treated with 5 mg/kg firocoxib administered orally, once a day for 52 weeks. Veterinary examinations were performed on approximately days 0, 15, 90, 180, 270 and 360. Twenty-five dogs completed the study. The withdrawal
Lauretta Maggi et al.
International journal of molecular sciences, 20(12) (2019-06-27)
An electrospinning process was optimized to produce fibers of micrometric size with different combinations of polymeric and surfactant materials to promote the dissolution rate of an insoluble drug: firocoxib. Scanning Electron Microscopy (SEM) showed that only some combinations of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service