Skip to Content
Merck
All Photos(2)

Documents

32094

Supelco

Penoxsulam

PESTANAL®, analytical standard

Synonym(s):

2-(2,2-Difluoroethoxy)-N-(5,8-dimethoxy[1,2,4]triazolo[1,5-c]pyrimidin-2-yl)-6-(trifluoromethyl)benzenesulfonamide, 3-(2,2-Difluoroethoxy)-N-(5,8-dimethoxy[1,2,4]triazolo[1,5-c]pyrimidin-2-yl)-α,α,α-trifluorotoluene-2-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14F5N5O5S
CAS Number:
Molecular Weight:
483.37
Beilstein:
10301178
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

SMILES string

COc1cnc(OC)n2nc(NS(=O)(=O)c3c(OCC(F)F)cccc3C(F)(F)F)nc12

InChI

1S/C16H14F5N5O5S/c1-29-10-6-22-15(30-2)26-13(10)23-14(24-26)25-32(27,28)12-8(16(19,20)21)4-3-5-9(12)31-7-11(17)18/h3-6,11H,7H2,1-2H3,(H,24,25)

InChI key

SYJGKVOENHZYMQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Penoxsulam is a triazolopyrimidine sulfonamide type of post-emergence herbicide, used as an acetolactate synthase (ALS) inhibitor for weed control management.

Application

Penoxsulam may be used as a reference standard in the determination of penoxsulam in:
  • Water samples using high performance liquid chromatography coupled with diode array detector (HPLC-DAD).
  • Aquatic systems adjacent to rice fields using liquid chromatography coupled with ultraviolet detector.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Development and application of an off-line SPE?LC?UV methodology for the determination of penoxsulam in aquatic systems adjacent to rice fields.
Cachada A.
Chromatographia, 71(3-4), 347-350 (2010)
Partitioning of penoxsulam, a new sulfonamide herbicide
Jabusch.WT, et al.
Journal of Agricultural and Food Chemistry, 53(18), 7179-7183 (2005)
Development and validation of an HPLC-DAD method for the simultaneous determination of most common rice pesticides in paddy water systems
Tsochatzis.DE, et al.
International Journal of Environmental Analytical Chemistry, 92(5), 548-560 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service