Skip to Content
Merck

Skip To

32043BC

VIP® Gold Salmonella Positive Control

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.85
UNSPSC Code:
41116129
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


shelf life

24 mo.

Quality Segment

technique(s)

lateral flow assay: suitable

application(s)

environmental
food and beverages
pathogen testing

compatibility

for use with BioControl VIP® Gold

storage temp.

2-8°C

General description

VIP® Gold Salmonella Positive Control, 5mL

Application

Lateral Flow

Other Notes

One bottle, 5.0ml

Legal Information

VIP is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
35044BC60038BC34761BC
compatibility

for use with BioControl VIP® Gold

compatibility

for use with BioControl VIP® Gold

compatibility

-

compatibility

for use with BioControl GDS

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

room temp

storage temp.

2-8°C

Quality Level

100

Quality Level

100

Quality Level

-

Quality Level

100

application(s)

environmental
food and beverages
pathogen testing

application(s)

environmental
food and beverages

application(s)

food and beverages
pathogen testing

application(s)

food and beverages
pathogen testing

shelf life

24 mo.

shelf life

22 mo.

shelf life

3‑9 mo.

shelf life

12 mo.

technique(s)

lateral flow assay: suitable

technique(s)

lateral flow assay: suitable

technique(s)

lateral flow assay: suitable, microbe id | immunological: suitable

technique(s)

PCR: suitable


hcodes

Hazard Classifications

Aquatic Chronic 3

Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters