Skip to Content
Merck
All Photos(2)

Documents

31102

Supelco

2,6-Dichlorophenol

PESTANAL®, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cl2C6H3OH
CAS Number:
Molecular Weight:
163.00
Beilstein:
1447806
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

bp

218-220 °C (lit.)

mp

64-66 °C (lit.)

application(s)

agriculture
environmental

format

neat

SMILES string

Oc1c(Cl)cccc1Cl

InChI

1S/C6H4Cl2O/c7-4-2-1-3-5(8)6(4)9/h1-3,9H

InChI key

HOLHYSJJBXSLMV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dichlorophenol is an organochloride of phenol and also a degradation product of some pesticides.Chloride phenols are widely used by the chemical industry as an intermediate product in synthesis and previously were frequently applied as wood preservatives and fungicides. They are considered one of the emerging pollutants in wastewater due to their extreme corrosive nature and toxicity even at low concentrations.

Application

2,6-Dichlorophenol may have the following uses:
  • To study the kinetics, performance, and mechanism of the oxidative degradation of 2,6-dichlorophenol (2,6-DCP) by ferrate (VI) (Fe(VI))
  • Removal of 2,6-dichlorophenol in water by copper oxide (CuO) activated peroxymonosulfate as catalyst
  • Removal of 2,6-dichlorophenol by adsorption with activated polypropylene nanofiber
  • Degradation of 2,6-dichlorophenol by Fe-doped titanium oxide(TiO2) sonophotocatalytic process
  • Determination of 2,6-dichlorophenol by surface-enhanced Raman scattering (SERS) with molecular imprinting

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lígia Miranda Ferreira Borges et al.
Veterinary parasitology, 147(1-2), 155-160 (2007-04-27)
This study was carried out with the objective of evaluating the efficacy of a 2,6-dichlorophenol (2,6-DCP) lure to control Dermacentor nitens (Acari: Ixodidae). Slow-release formulations of the pheromone formulated with and without cypermethrin were prepared. Olfactometer bioassays were used to
Davide Vione et al.
Chemosphere, 69(10), 1548-1554 (2007-07-10)
2,4-Dichlorophenol (2,4-DCP) and 2,6-dichlorophenol (2,6-DCP) undergo oxidation, nitrosation and nitration in the presence of nitrate under UV irradiation. Nitration is favoured under acidic conditions, most likely because HNO(2) is formed on nitrate photolysis. The most likely photonitration pathway is the
Yu Tian et al.
Water research, 47(1), 279-288 (2012-11-06)
Distribution variation of a metabolic uncoupler, 2,6-dichlorophenol (2,6-DCP), in long-term sludge culture was studied, and the effects on sludge reduction and biological inhibition of this chemical during the 90-day operation were established. The extracellular polymeric substance (EPS) matrix functioned as
Qiang He et al.
Applied microbiology and biotechnology, 65(4), 377-382 (2004-06-05)
A continuous fed-batch reactor system was developed to rapidly obtain dense chlororespiring cultures of Anaeromyxobacter dehalogenans strain 2CP-C. A syringe pump continuously delivered concentrated 2,6-dichlorophenol (50-150 mM) to an anaerobic reactor vessel at a rate that sustained linear growth but
S Almeda et al.
Analytica chimica acta, 587(1), 97-103 (2007-03-28)
Actually there is a great trend on the development of effective analytical methods for monitoring trace levels of various phenols which can indicate, among others compounds, the water quality. A simple, inexpensive supported liquid membrane (SLM) device was used in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service