Skip to Content
Merck
All Photos(1)

Documents

237868

Sigma-Aldrich

Brucine sulfate heptahydrate

ACS reagent

Synonym(s):

2,3-Dimethoxystrychnidin-10-one sulfate heptahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C23H26N2O4)2 · H2SO4 · 7H2O
CAS Number:
Molecular Weight:
1013.11
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

grade

ACS reagent

Quality Level

form

solid

optical activity

[α]22/D −24°, c = 1 in H2O

ign. residue

≤0.1%

loss

≤13.0% loss on drying

mp

180 °C (dec.) (lit.)

SMILES string

[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].[H]O[H].OS(O)(=O)=O.[H][C@@]12CC(=O)N3c4cc(OC)c(OC)cc4[C@]56CC[N@@H]7CC(=CCO1)[C@H](C[C@@H]57)[C@]2([H])[C@]36[H].[H][C@@]89CC(=O)N%10c%11cc(OC)c(OC)cc%11[C@]%12%13CC[N@@H]%14CC(=CCO8)[C@H](C[C@@H]%12%14)[C@]9([H])[C@]%10%13[H]

InChI

1S/2C23H26N2O4.H2O4S.7H2O/c2*1-27-16-8-14-15(9-17(16)28-2)25-20(26)10-18-21-13-7-19-23(14,22(21)25)4-5-24(19)11-12(13)3-6-29-18;1-5(2,3)4;;;;;;;/h2*3,8-9,13,18-19,21-22H,4-7,10-11H2,1-2H3;(H2,1,2,3,4);7*1H2/t2*13-,18-,19-,21-,22-,23+;;;;;;;;/m00......../s1

InChI key

PPJIVFYMRNHHTJ-JYGVWXPPSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 3

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service