Skip to Content
Merck

23258

2,2,2-Trichloroethyl chloroformate

purum, ≥98.0% (NT)

Synonym(s):

2,2,2-Trichloroethoxycarbonyl chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
ClCOOCH2CCl3
CAS Number:
Molecular Weight:
211.86
UNSPSC Code:
12352100
NACRES:
NA.22
PubChem Substance ID:
EC Number:
241-363-7
Beilstein/REAXYS Number:
970619
MDL number:
Assay:
≥98.0% (NT)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


vapor pressure

0.06 psi ( 20 °C)

Quality Segment

grade

purum

assay

≥98.0% (NT)

refractive index

n20/D 1.471

bp

171-172 °C (lit.)

density

1.539 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC(=O)OCC(Cl)(Cl)Cl

InChI

1S/C3H2Cl4O2/c4-2(8)9-1-3(5,6)7/h1H2

InChI key

LJCZNYWLQZZIOS-UHFFFAOYSA-N

Application

2,2,2-Trichloroethyl chloroformate was used as:
  • derivatizing reagent in gas chromatographic/mass spectrometric determination of a large range of amphetamine-related drugs and ephedrines in plasma, urine and hair samples
  • starting reagent during the synthesis of 6-nor-9,10-dihydrolysergic acid methyl ester (IV)
  • reagent during N-demethylation of dextromethorphan
  • protecting group reagent for amines and alcohols


Still not finding the right product?

Explore all of our products under 2,2,2-Trichloroethyl chloroformate


Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 3 Inhalation - Eye Dam. 1 - Skin Corr. 1B

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Faceshields, Gloves, Goggles, type ABEK (EN14387) respirator filter



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library