Skip to Content
Merck

217441

AmberChrom 50WX2 Ion Exchange Resin

hydrogen form, 50-100 mesh, cationic

Synonym(s):

AmberChrom 50WX2 hydrogen form

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
23151817
NACRES:
SB.52
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

AmberChrom 50WX2 50-100 Mesh (H+) Cation Exchange Resin, hydrogen form, 50-100 mesh

description

cross-linkage 2%

Quality Segment

form

beads

parameter

150 °C max. temp.

moisture

74-82%

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

matrix active group

sulfonic acid functional group

particle size

50-100 mesh

operating pH

0-14

capacity

0.6 meq/mL by wetted bed volume

separation technique

cation exchange

SMILES string

C(C)c3c(cccc3)C=C.c2(c(cccc2)C=C)C=C.c1(ccccc1)C=C

InChI

1S/C10H12.C10H10.C8H8/c2*1-3-9-7-5-6-8-10(9)4-2;1-2-8-6-4-3-5-7-8/h3,5-8H,1,4H2,2H3;3-8H,1-2H2;2-7H,1H2

InChI key

NWUYHJFMYQTDRP-UHFFFAOYSA-N

General description

AmberChrom 50WX2 50-100 Mesh (H+) Cation Exchange Resin is a heat stable, water stable and low molecular weight cationic substance which is capable of purifying approximately 5000-fold by ion-exchange chromatography. Ion-exchange resins have good role as drug delivery systems.
Strongly acidic cation exchange resin. This ion-exchange resin has not been specially processed or cleaned. We suggest that it be subjected to a suitable preliminary elution and wash prior to use.

Application

AmberChrom 50WX2 50-100 Mesh (H+) Cation Exchange Resinwas used to remove calcium from aqueous samples.

Legal Information

AmberChrom is a trademark or registered trademark of affiliates of DuPont de Nemours, Inc.
Amberchrom is a trademark of DuPont de Nemours, Inc.


Still not finding the right product?


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Eye Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library