Skip to Content
Merck
All Photos(2)

Key Documents

215597

Sigma-Aldrich

Basic Fuchsin

Dye content >85 %

Synonym(s):

Basic Parafuchsin, Basic Red 9, Magenta O, Parafuchsin hydrochloride, Paramagenta hydrochloride, Pararosaniline chloride, Pararosaniline hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H17N3 · HCl
CAS Number:
Molecular Weight:
323.82
Colour Index Number:
42500
Beilstein:
4164603
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

Assay

>85.0% (HPLC)

form

solid

composition

Dye content, >85%

technique(s)

microbe id | staining: suitable

mp

268-270 °C (dec.) (lit.)

λmax

545 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Nc1ccc(cc1)C(\c2ccc(N)cc2)=C3/C=CC(=[NH2+])C=C3

InChI

1S/C19H17N3.ClH/c20-16-7-1-13(2-8-16)19(14-3-9-17(21)10-4-14)15-5-11-18(22)12-6-15;/h1-12,20H,21-22H2;1H

InChI key

JUQPZRLQQYSMEQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Basic fuchsin is a triaminotriphenylmethane dye that is widely used to stain muscle, mitochondria, and collagen. It is generally a mixture of pararosaniline (C.I. 42500), rosaniline (C.I. 42510), magenta II and new fuchsine (CI 42520).

Application

Basic fuchsin is utilized for various applications:
  • Acid-fast staining
  • Periodic Acid-Schiff
  • Feulgen stain for DNA
  • ENDO culture medium
  • Leifson′s method for bacterial flagella

Biochem/physiol Actions

Basic fuchsin displays anesthetic, bactericidal (gram-positive), and fungicidal properties.

Legal Information

Magenta is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design and Fabrication of Standardized Hydroxyapatite Scaffolds with a Defined Macro-Architecture by Rapid Prototyping for Bone-Tissue-Engineering Research
Wilson CE
Journal of Biomedical Materials Research Part A, 68, 123-132 (2004)
Archarna Sharma
Plant Chromosomes (1999)
Hans-Ferdinand Linskens, John F. Jackson
Plant Fibers, 10 (2012)
Bisen PS
Laboratory Protocols in Applied Life Sciences (2014)
Luis F Maceda-López et al.
3 Biotech, 11(2), 75-75 (2021-01-29)
Together with their undeniable role in the ecology of arid and semiarid ecosystems, Agave species are emerging as a model to dissect the relationships between crassulacean acid metabolism and high efficiency of light and water use, and as an energy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service