Skip to Content
Merck
All Photos(1)

Key Documents

19667

Supelco

Butylboronic acid

for GC derivatization, LiChropur, ≥96.0% (T)

Synonym(s):

1-Butaneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3B(OH)2
CAS Number:
Molecular Weight:
101.94
Beilstein:
1733489
EC Number:
MDL number:
UNSPSC Code:
41116105
PubChem Substance ID:
NACRES:
NA.22

grade

for GC derivatization

Quality Level

Assay

≥96.0% (T)

quality

LiChropur

reaction suitability

reagent type: derivatization reagent
reaction type: Alkylations

technique(s)

gas chromatography (GC): suitable

mp

90-92 °C (lit.)
90-92 °C

SMILES string

CCCCB(O)O

InChI

1S/C4H11BO2/c1-2-3-4-5(6)7/h6-7H,2-4H2,1H3

InChI key

QPKFVRWIISEVCW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Developing a reference measurement procedure for free glycerol in human serum by two-step gas chromatography-isotope dilution mass spectrometry.: This research employs butylboronic acid in a reference measurement procedure to quantify free glycerol in human serum. The method is significant for clinical diagnostics, providing accurate and reliable measurements for metabolic studies (Chen et al., 2015).

Other Notes

Derivatizing agent for gas chromatography
Reagent for butylboronate.
Suitable for the derivatization of proximal difunctional compounds (a- and ß-hydroxyacids; Ο-phenolic acids; enolizable a-oxo acids; 1,2- and 1,3-diols; enolizable 1,2- and 1,3-ketols; ß- and γ-hydroxyamines).

Legal Information

LiChropur is a trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

D.R. Knapp
Handbook of Analytical Derivatization Reactions (1979)
A. Darbre et al.
Handbook of Derivatives for Chromatography (1970)
[Gas chromatographic separation of the diastereomers of labetalol as n-butylboronate].
T Goromaru et al.
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 103(9), 974-978 (1983-09-01)
F Ramos et al.
Journal of chromatography. B, Biomedical sciences and applications, 716(1-2), 366-370 (1998-11-21)
A derivatization procedure for confirmatory residue analysis of beta2-agonists is described. Methyl (MBA) and butyl (BBA) boronic acids are simultaneously used for the derivatization of tulobuterol, mabuterol, mapenterol, salbutamol, clenproperol, clenbuterol, clenpenterol and bromobuterol by GC-MS determination. A temperature of
M Inouye et al.
Japanese journal of medicine, 27(1), 29-33 (1988-02-01)
A highly sensitive and specific quantitative assay for inositol in serum has been developed. Uremic serum, to which hexahydroxybenzene as an internal standard was added, was deproteined with trichloroacetate. The supernatant aqueous phase was taken after lipids in serum were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service