Skip to Content
Merck
All Photos(1)

Documents

19600

Supelco

Butylbenzene

analytical standard

Synonym(s):

1-Phenylbutane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)3CH3
CAS Number:
Molecular Weight:
134.22
Beilstein:
1903395
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

vapor density

>1 (vs air)

vapor pressure

1.03 mmHg ( 23 °C)

Assay

≥99.8% (GC)

autoignition temp.

774 °F

shelf life

limited shelf life, expiry date on the label

expl. lim.

5.8 %

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.489 (lit.)
n20/D 1.490

bp

183 °C (lit.)

mp

−88 °C (lit.)

density

0.86 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CCCCc1ccccc1

InChI

1S/C10H14/c1-2-3-7-10-8-5-4-6-9-10/h4-6,8-9H,2-3,7H2,1H3

InChI key

OCKPCBLVNKHBMX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Butylbenzene is a colourless organic solvent. It is used in pesticide manufacturing, plasticizer and also as solvent for coating. It is sometimes used as surface active agent, polymer linking agent and in organic synthesis.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

138.2 °F - closed cup

Flash Point(C)

59.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Technology and Engineering
Groundwater Chemicals Desk Reference, 177-178 (2010)
Technology and Engineering
Montgomery, John H.
Groundwater Chemicals Desk Reference, 14-Dec, 177-178 (2010)
M Takeshita et al.
Research communications in molecular pathology and pharmacology, 89(3), 351-356 (1995-09-01)
Regio- and stereo-selective oxidation of butylbenzene (1) has been examined in vitro by rat liver supernatant fraction(S-9). When phenylbutane (1 ) was incubated at 37 degrees C for 1 hr with S-9, asymmetric oxidation occurred regioselectively at benzylic and omega-1
Stéphanie Halbert et al.
Journal of computational chemistry, 32(8), 1550-1560 (2011-02-18)
Conformational landscape of neutral and ionized n-butylbenzene has been examined. Geometries have been optimized at the B3LYP/6-31G(d), B3LYP/6-31+G(d,p), B3LYP-D/6-31+G(d,p), B2PLYP/6-31+G(d,p), B2PLYP-D/6-31+G(d,p), B97-D/6-31+G(d,p), and M06-2X/6-31+G(d,p) levels. This study is complemented by energy computations using 6-311++G(3df,2p) basis set and CBS-QB3 and G3MP2B3
Hiroyuki Izumi et al.
The Journal of toxicological sciences, 30 Spec No., 21-38 (2006-04-28)
N-butylbenzene (n-BB), which is suspected of having endocrine disrupting effects, was administered orally by gavage at dose levels of 0, 30, 100, and 300 mg/kg/day to groups of Crj: CD (SD) IGS rats (24 males and 24 females per group)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service