Skip to Content
Merck

Skip To

17779

2,3,5-Triphenyl-tetrazolium chloride solution

suitable for microbiology, Filter sterilized solution that is recommended for the detection of microbial growth based on reduction of TTC

Synonym(s):

TTC solution, TTC supplement, TTC solution, TTC supplement

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C19H15ClN4
CAS Number:
Molecular Weight:
334.80
UNSPSC Code:
41171614
NACRES:
NA.85
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
3923356
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

sterility

sterile (Filtered and Aseptic Handled)

form

liquid

shelf life

limited shelf life, expiry date on the label

mol wt

334.8 g/mol

application(s)

agriculture
environmental
food and beverages, microbiology

storage temp.

2-8°C

suitability

bacteria

SMILES string

[Cl-].c1ccc(cc1)-c2nn(-c3ccccc3)[n+](n2)-c4ccccc4

InChI

1S/C19H15N4.ClH/c1-4-10-16(11-5-1)19-20-22(17-12-6-2-7-13-17)23(21-19)18-14-8-3-9-15-18;/h1-15H;1H/q+1;/p-1

InChI key

PKDBCJSWQUOKDO-UHFFFAOYSA-M

General description

2,3,5-Triphenyl-tetrazolium chloride (TTC) is a dye largely used for the easy enumeration of microbial colonies in solid culture media. This dye is colorless in the oxidized form and changes to red when reduced by microorganisms, due to the formation of formazan. Live microorganisms, for example, E. coli and other coliforms reduce TTC by enzymatic action, forming red color formazan that is kept inside granules in the cells. TTC is highly recommended in testing pasteurized milk because there is a high percentage of microorganisms unable to reduce TTC in pasteurized milk, which cannot be detected by laboratory procedures. Higher concentrations of TTC may have an inhibitory effect on the growth of Gram-positive organisms.

Application

2,3,5-Triphenyl-tetrazolium chloride (TTC) solution is a redox indicator that can help indicate cellular respiration and identify viable and non-viable cells as well as identify microorganisms capable of reduction of TTC on solid culture media

Other Notes

(per vial)
2,3,5-Triphenyl tetrazolium chloride: 0.1 g, distilled water: 10 ml

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
54232T9656T7791
suitability

bacteria

suitability

coliforms, selective and differential for Citrobacter spp., selective and differential for Enterobacter spp., selective and differential for Escherichia coli, selective and differential for Proteus spp., selective and differential for Salmonella spp., selective and differential for Shigella spp.

suitability

-

suitability

-

application(s)

agriculture
environmental
food and beverages, microbiology

application(s)

agriculture
bioburden testing
environmental
food and beverages
water monitoring, microbiology

application(s)

-

application(s)

-

form

liquid

form

powder

form

liquid

form

liquid

sterility

sterile (Filtered and Aseptic Handled)

sterility

non-sterile

sterility

-

sterility

-

storage temp.

2-8°C

storage temp.

-

storage temp.

−20°C

storage temp.

−20°C

shelf life

limited shelf life, expiry date on the label

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters