Skip to Content
Merck
All Photos(3)

Key Documents

157775

Sigma-Aldrich

Phosphorus pentachloride

reagent grade, 95%

Synonym(s):

Phosphorus(V) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PCl5
CAS Number:
Molecular Weight:
208.24
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.21
grade:
reagent grade
form:
powder

grade

reagent grade

Quality Level

vapor pressure

<1 mmHg ( 20 °C)

Assay

95%

form

powder

pH

1 (5 g/L)

mp

179-181 °C (subl.) (lit.)

SMILES string

ClP(Cl)(Cl)(Cl)Cl

InChI

1S/Cl5P/c1-6(2,3,4)5

InChI key

UHZYTMXLRWXGPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phosphorus pentachloride is a light yellow colored phosphorous halide. It participates in the synthesis of thiazoline analogs.

Application

Phosphorus pentachloride may be used in the synthesis of following compounds:
  • benzenesulfonyl chloride
  • malonic dinitrile
  • aromatic sulfonyl chlorides

Phosphorus pentachloride may be used as halogenating reagent to convert any substituted aldehyde into the corresponding vinyl halide. It can also undergo reaction with ammonium chloride to yield linear phosphazenes and cyclophosphazenes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Allcock H
Phosphorus-Nitrogen Compounds: Cyclic, Linear, and High Polymeric Systems (2012)
Ley.VS et al.
Comprehensive Organic Functional Group Transformations: Synthesis: carbon with two heteroatoms, each attached by a single bond (2005)
Ley.VS
Comprehensive Organic Functional Group Transformations II, 2 (1995)
Synthetic studies of thiazoline and thiazolidine-containing natural products-1. Phosphorus pentachloride-mediated thiazoline construction reaction.
Ino A and Murabayashi A.
Tetrahedron, 55(34), 10271-10282 (1999)
Eagleson M.
Concise Encyclopedia Chemistry, 120-120 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service