156477
4-(Dimethylamino)benzaldehyde
ACS reagent, 99%
Synonym(s):
Ehrlich’s reagent
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2NC6H4CHO
CAS Number:
Molecular Weight:
149.19
Beilstein:
606802
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21
Recommended Products
grade
ACS reagent
Quality Level
Assay
99%
form
powder
ign. residue
≤0.1%
mp
72-75 °C (lit.)
solubility
alcohol: passes test (APHA ≤60)
hydrochloric acid: passes test
λmin
369 nm
λmax
385 nm
functional group
aldehyde
amine
SMILES string
CN(C)c1ccc(C=O)cc1
InChI
1S/C9H11NO/c1-10(2)9-5-3-8(7-11)4-6-9/h3-7H,1-2H3
InChI key
BGNGWHSBYQYVRX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
4-(Dimethylamino)benzaldehyde has been used in the preparation of trans-4-dimethylamino-4′-nitrostilbene.
Forms colored condensation products (Schiff bases) with pyrroles and primary amines.
Forms colored condensation products (Schiff bases) with pyrroles and primary amines.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Sens. 1B
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
327.2 °F - closed cup
Flash Point(C)
164 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Na Cao et al.
Nature communications, 10(1), 2877-2877 (2019-06-30)
The electrochemical N2 fixation to produce ammonia is attractive but significantly challenging with low yield and poor selectivity. Herein, we first used density function theory calculations to reveal adjacent bi-Ti3+ pairs formed on anatase TiO2 as the most active electrocatalytic
The Journal of Organic Chemistry, 59, 6038-6038 (1994)
A protocol for preparing, characterizing and using three RNA-specific, live cell imaging probes: E36, E144 and F22
Li Q & Chang YT
Nature Protocols, 1(6), 2922-2922 (2006)
Vladislav Papper et al.
Journal of fluorescence, 28(1), 13-19 (2014-08-12)
N,N-dimethyl-N'-picryl-4,4'-stilbenediamine (DMPSDA) was prepared, purified and crystallised in a form of black lustrous crystals, and its absorption and fluorescence spectra were recorded in cyclohexane, acetonitrile and dimethyl sulfoxide. Non-emissive intramolecular charge transfer state (ICT) was clearly observed in this molecule
Excited and ground state conformations of p-dimethylamino-benzaldehyde and p-dimethylaminoacetophenone.
Dobkowski J, et al.
Journal of Luminescence, 27(4), 339-355 (1982)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service