Skip to Content
Merck
All Photos(2)

Key Documents

15598

Supelco

(−)-Borneol

analytical standard

Synonym(s):

endo-(1S)-1,7,7-Trimethylbicyclo[2.2.1]heptan-2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2038053
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥99.0% (sum of enantiomers, GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

food and beverages

format

neat

SMILES string

CC1(C)[C@H]2CC[C@]1(C)[C@H](O)C2

InChI

1S/C10H18O/c1-9(2)7-4-5-10(9,3)8(11)6-7/h7-8,11H,4-6H2,1-3H3/t7-,8+,10+/m0/s1

InChI key

DTGKSKDOIYIVQL-QXFUBDJGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

(-)-Borneol is an enantiomer. It is a bicyclic monoterpene compound used gengrally for analgesia and anaesthesia. It is considered as positive modulators of GABA receptors.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Sol. 2 - Skin Sens. 1

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 2

Flash Point(F)

150.8 °F

Flash Point(C)

66 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

"(+)-And (-)-borneol: efficacious positive modulators of GABA action at human recombinant a 1 ? 2 ? 2L GABA A receptors.
Granger, Renee E., Erica L. Campbell, and Graham AR Johnston.
Biochemical Pharmacology, 69.7, 1101-1111 (2005)
Eva Horváthová et al.
Mutagenesis, 27(5), 581-588 (2012-05-01)
Experimental evidences suggest that most essential oils possess a wide range of biological and pharmacological activities that may protect tissues against oxidative damage. In this study, we investigated DNA-protective effect of borneol, a component of many essential oils, against oxidative
Xiao-Hong Wei et al.
Microcirculation (New York, N.Y. : 1994), 20(1), 17-29 (2012-08-24)
The present study was designed to evaluate whether CP was beneficial in alleviating myocardial fibrosis following I/R injury. Sprague-Dawley rats were subjected to 30 minutes occlusion of the LADCA, followed by reperfusion. CP (0.4 or 0.8 g/kg) was daily administered starting from
Jing-yi Zhao et al.
Journal of Zhejiang University. Science. B, 13(12), 990-996 (2012-12-12)
Borneol, a monoterpenoid alcohol, is used widely, particularly in combined formulas for preventing and curing cardiovascular and cerebrovascular diseases in traditional Chinese medicine. In order to understand the blood and brain pharmacokinetics after intravenous, intranasal, or oral administration and to
Jingjing Liu et al.
European journal of pharmacology, 688(1-3), 1-5 (2011-12-28)
The bioavailabilities of eye drops are very low, so it is eager to find a safe and effective penetration enhancer to improve drug bioavailability. In our study, the corneas of New Zealand albino rabbit were mounted in the improved Franz

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service