Skip to Content
Merck
All Photos(2)

Key Documents

15343

Supelco

Lead ionophore IV

Selectophore

Synonym(s):

tert-Butylcalix[4]arene-tetrakis(N,N-dimethylthioacetamide)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C60H84N4O4S4
CAS Number:
Molecular Weight:
1053.59
MDL number:
UNSPSC Code:
26111700
NACRES:
NB.61

grade

for ion-selective electrodes

Quality Level

product line

Selectophore

General description

Visit our Sensor Applications portal to learn more.

Application

Lead ionophore IV (tert-Butylcalix[4]arene-tetrakis(N,N-dimethylthioacetamide)), was used in the fabrication of robust potentiometric ion sensors. It was also used as the base for the synthesis of lead-selective micro-electrode.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Ionophore giving highly Pb-selective polymer membrane electrodes

Legal Information

Selectophore is a trademark of Merck KGaA, Darmstadt, Germany

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Evaluation of Liquid- and Solid-Contact, Pb2+-Selective Polymer-Membrane Electrodes for Soil Analysis
McGraw, C. M., et al.
Electroanalysis, 20 (3), 340-346 (2008)
E. Malinowska et al.
Analytica Chimica Acta, 298, 253-253 (1994)
A solid-contact Pb 2+-selective electrode using poly (2-methoxy-5-(2'-ethylhexyloxy)-p-phenylene vinylene) as ion-to-electron transducer.
Shunyang Y
Analytica Chimica Acta, 702 (2), 195-198 (2011)
New Solid-State Organic Membrane Based Lead-Selective Micro-Electrode.
Arida H
International Journal of Electrochemical Science, 6 (9), 3858-3867 (2011)
Chantana Bualom et al.
Talanta, 82(2), 660-667 (2010-07-07)
A sensitive optode consisting of highly lead-selective ionophore (Lead IV), proton-selective chromoionophore (ETH 5294) and lipophilic anionic sites (KTpClPB) in plasticized polyvinyl chloride (PVC) membrane was fabricated. The optode membranes were used for determination of Pb(2+) by absorption spectrophotometry in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service