Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
CAS Number:
UNSPSC Code:
23201100
PubChem Substance ID:
NACRES:
SB.52
EC Number:
231-545-4
MDL number:
Product Name
Silica gel orange, granular, 0.2-1 mm
grade
high-purity grade
Quality Segment
form
granular
technique(s)
LPLC: suitable
surface area
500 m2/g , approx.
matrix
Silica
matrix active group
silica
particle size
0.2-1 mm
pore size
60 Å mean pore size
bp
2230 °C
mp
>1600 °C
separation technique
hydrophilic interaction (HILIC)
SMILES string
O=[Si]=O
InChI
1S/O2Si/c1-3-2
InChI key
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
General description
Silica gel, highly porous textured matrix are generally used for synthesis of adsorbents. They are corpuscular in structure. Silica surface have a layer of hydroxyl groups and adsorbed water molecules are removed by drying at high temperature or even by treatment with suitable reagents. Silica gel surface plays significant role in catalysis, chemical separation and microelectronic fabrications.
Still not finding the right product?
Explore all of our products under Silica gel orange
Storage Class
11 - Combustible Solids
WGK
nwg
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
PPE (personal protective equipment)
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.