Sign In to View Organizational & Contract Pricing.
Select a Size
About This Item
NACRES:
NA.24
UNSPSC Code:
41116107
grade
analytical standard
manufacturer/tradename
Whitehouse Scientific Ltd PS315
technique(s)
diffraction/scattering: suitable
particle size
10-100 μm
application(s)
cosmetics
environmental
food and beverages
pharmaceutical
format
neat
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Polydisperse Particle Standard (PS315, size: 10-100 μm) is appropriate to assess the particle size distribution (PSD) profile of a particle system.
Application
Used to validate and calibrate particle size equipment.
It can also be used to test and monitor the precision and reproducibility of laser diffraction particle analyzer.
It can also be used to test and monitor the precision and reproducibility of laser diffraction particle analyzer.
Features and Benefits
- suitable for routine instrument calibration checks, testing and corrections
- available as a neat sample
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Soil variables that determine lead accumulation in Bidens pilosa L. and Tagetes minuta L. growing in polluted soils.
Salazar M J, et al.
Geoderma, 279, 97-108 (2016)
Revisiting the chronology and environmental conditions for the accretion of late Pleistocene-early Holocene Pampean loess (Argentina)
Torre G, et al.
Chemosphere, 213, 105-119 (2019)
Arsenic-bearing phases in South Andean volcanic ashes: Implications for As mobility in aquatic environments
Bia G, et al.
Chemical Geology , 393, 26-35 (2015)
The paleoclimatic message from the polymodal grain-size distribution of late Pleistocene-early Holocene Pampean loess (Argentina)
Torre G, et al.
Chemosphere, 42, 100563-100563 (2020)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 525200-4L | 04061838351470 |
| 525200-20L | 04061832550466 |
| 525200-500ML | 04061832551876 |
| 08718-10X1G | 04061833387573 |
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service