Skip to Content
Merck

Skip To

08421

Benzyl alcohol

analytical standard

Synonym(s):

Benzenemethanol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
C6H5CH2OH
CAS Number:
Molecular Weight:
108.14
NACRES:
NA.24
PubChem Substance ID:
eCl@ss:
39023110
UNSPSC Code:
12352104
EC Number:
202-859-9
MDL number:
Beilstein/REAXYS Number:
878307
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

vapor density

3.7 (vs air)

vapor pressure

13.3 mmHg ( 100 °C), 3.75 mmHg ( 77 °C)

assay

≥99.5% (GC)

autoignition temp.

817 °F

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

refractive index

n20/D 1.539 (lit.), n20/D 1.541

bp

203-205 °C (lit.)

mp

−16-−13 °C (lit.)

density

1.045 g/mL at 25 °C (lit.)

application(s)

environmental
food and beverages

format

neat

SMILES string

OCc1ccccc1

InChI

1S/C7H8O/c8-6-7-4-2-1-3-5-7/h1-5,8H,6H2

InChI key

WVDDGKGOMKODPV-UHFFFAOYSA-N

General description

Benzyl alcohol belongs to the aromatic alcohol group of compounds widely utilized in cosmetics. It is also used as a solvent and viscosity-reducing agent.
This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food. Find all available reference materials for compounds listed in 10/2011 here

Application

Benzyl alcohol may be used as an analytical standard for the determination of benzyl alcohol in:
  • Wine samples by dichloromethane microextraction followed by gas chromatography-flame ionization detection (GC-FID).
  • Uncased and cased tobaccos by stream distillation continuous extraction (SDE) followed by capillary GC or GC coupled with mass spectroscopy (MS).
  • Dairy products by solid phase microextraction (SPME) followed by GC-MS.
  • Scented products by reversed phase high performance liquid chromatography (HPLC) combined with diode array detector (DAD).

Other Notes

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.
This compound is commonly found in plants of the genus: ginkgo

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR1019W213713W213705
description

analytical standard

description

Pharmaceutical Secondary Standard; Certified Reference Material

description

natural, ≥98%, FG

description

≥99%, FCC, FG

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

UV/Vis spectroscopy: suitable

technique(s)

-

grade

analytical standard

grade

certified reference material, pharmaceutical secondary standard

grade

FG, Fragrance grade, Halal, Kosher, natural

grade

FG, Halal, Kosher

application(s)

environmental
food and beverages

application(s)

pharmaceutical (small molecule)

application(s)

flavors and fragrances

application(s)

flavors and fragrances

format

neat

format

neat

format

-

format

-

assay

≥99.5% (GC)

assay

-

assay

≥98%

assay

≥99%

bp

203-205 °C (lit.)

bp

203-205 °C (lit.)

bp

203-205 °C (lit.)

bp

203-205 °C (lit.)


Still not finding the right product?

Explore all of our products under Benzyl alcohol


pictograms

Exclamation mark

signalword

Warning

Storage Class

10 - Combustible liquids

flash_point_f

213.8 °F - DIN 51758

flash_point_c

101 °C - DIN 51758

ppe

Eyeshields, Faceshields, Gloves, type ABEK (EN14387) respirator filter

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1B



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–2 of 2 Questions  
  1. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/418/501/product-dating-information-06-25-mk.pdf

      Helpful?

  2. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters