Skip to Content
Merck

Skip To

08066

Oleyl alcohol

analytical standard

Synonym(s):

Z-9-Dodecen-1-ol, cis-9-Octadecen-1-ol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
CH3(CH2)7CH=CH(CH2)7CH2OH
CAS Number:
Molecular Weight:
268.48
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
EC Number:
205-597-3
Beilstein/REAXYS Number:
1723962
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥90.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤0.5% water

refractive index

n20/D 1.457-1.462, n20/D 1.46 (lit.)

bp

207 °C/13 mmHg (lit.)

mp

0-5.0 °C (lit.)

density

0.849 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

format

neat

storage temp.

−20°C

SMILES string

CCCCCCCC\C=C/CCCCCCCCO

InChI

1S/C18H36O/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19/h9-10,19H,2-8,11-18H2,1H3/b10-9-

InChI key

ALSTYHKOOCGGFT-KTKRTIGZSA-N

General description

This substance is listed on the positive list of the EU regulation 10/2011 for plastics intended to come into contact with food.

Find all available reference materials for compounds listed in 10/2011 here

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
O8880369314PHR2637
application(s)

cleaning products
cosmetics
flavors and fragrances
food and beverages
personal care

application(s)

-

application(s)

-

application(s)

pharmaceutical

grade

analytical standard

grade

-

grade

technical grade

grade

certified reference material, pharmaceutical secondary standard

format

neat

format

-

format

-

format

-

assay

≥90.0% (GC)

assay

≥99% (GC)

assay

85%

assay

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

mp

0-5.0 °C (lit.)

mp

0-5.0 °C (lit.)

mp

0-5.0 °C (lit.)

mp

0-5.0 °C (lit.)


Still not finding the right product?

Explore all of our products under Oleyl alcohol


Storage Class

10 - Combustible liquids

wgk

awg

flash_point_f

338.0 °F - closed cup

flash_point_c

170 °C - closed cup



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters