Skip to Content
Merck
All Photos(2)

Key Documents

07937

Supelco

Sudan II

analytical standard

Synonym(s):

1-(2,4-Xylylazo)-2-naphthol, Solvent Orange 7

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N2O
CAS Number:
Molecular Weight:
276.33
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥96.0% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

156-158 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

Cc1ccc(\N=N\c2c(O)ccc3ccccc23)c(C)c1

InChI

1S/C18H16N2O/c1-12-7-9-16(13(2)11-12)19-20-18-15-6-4-3-5-14(15)8-10-17(18)21/h3-11,21H,1-2H3/b20-19+

InChI key

JBTHDAVBDKKSRW-FMQUCBEESA-N

Looking for similar products? Visit Product Comparison Guide

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

448.9 °F - closed cup

Flash Point(C)

231.6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jia Wang et al.
Food chemistry, 136(3-4), 1478-1483 (2012-12-01)
A semiquantitative dip strip assay was developed using nanocolloidal gold-labelled monoclonal antibody (Mab) 8A10 for the rapid detection of Sudan red I in food samples. A protein-Sudan red I conjugate was coated on a nitro cellulose membrane strip in a
Zhiping Zhang et al.
The Analyst, 137(11), 2556-2558 (2012-04-26)
A simple and efficient ambient ionization method based on paper spray combined with tandem mass spectrometry allows rapid detection and quantitation of various contaminants (clenbuterol, melamine, plasticizer and sudan red) in various foodstuffs (e.g., meat, milk, sports drinks and chili
Claudio Baggiani et al.
Journal of separation science, 32(19), 3292-3300 (2009-09-02)
A method for molecularly imprinted SPE of banned Sudan azo-dyes from food samples was investigated. The molecularly imprinted polymer was obtained by suspension polymerization using 1-(4-chlorophenyl)azonaphthalen-2-ol as the mimic template. The molecular recognition properties of imprinted beads were evaluated for
Els Houben et al.
Analytical chemistry, 82(2), 601-607 (2009-12-17)
The use of stable isotope labeled substrates and subsequent analysis of urinary nitrate, forms a noninvasive test for evaluation of the in vivo NO metabolism. The present paper describes a new method for simultaneous quantification of (15)N-nitrate and total nitrate
Jing Xu et al.
The Analyst, 135(10), 2566-2572 (2010-08-07)
Sudan dyes are banned to be used in food additives because of the carcinogenicity of their metabolites. A rapid and sensitive indirect competitive enzyme-linked immunosorbent assay (ELISA) was developed to detect the residues of Sudan dyes. Novel immunogen and coating

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service