Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C8H12N2O3S
CAS Number:
Molecular Weight:
216.26
UNSPSC Code:
77101502
NACRES:
NA.24
PubChem Substance ID:
EC Number:
208-993-4
Beilstein/REAXYS Number:
15080
MDL number:
grade
analytical standard
Quality Level
assay
≥99.0% (HPLC)
optical activity
[α]/D 275±10°, c = 1 in 1 M HCl
shelf life
limited shelf life, expiry date on the label
technique(s)
HPLC: suitable, gas chromatography (GC): suitable
mp
198-200 °C (dec.) (lit.)
application(s)
pharmaceutical
format
neat
storage temp.
2-8°C
SMILES string
CC1(C)S[C@@H]2[C@H](N)C(=O)N2[C@H]1C(O)=O
InChI
1S/C8H12N2O3S/c1-8(2)4(7(12)13)10-5(11)3(9)6(10)14-8/h3-4,6H,9H2,1-2H3,(H,12,13)/t3-,4+,6-/m1/s1
InChI key
NGHVIOIJCVXTGV-ALEPSDHESA-N
Still not finding the right product?
Explore all of our products under (+)-6-Aminopenicillanic acid
signalword
Warning
hcodes
Hazard Classifications
Eye Irrit. 2
Storage Class
11 - Combustible Solids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Critical appraisal of pathology transmission in the α-synuclein fibril model of Lewy body disorders.
Negin Nouraei et al.
Experimental neurology, 299(Pt A), 172-196 (2017-10-24)
Lewy body disorders are characterized by the emergence of α-synucleinopathy in many parts of the central and peripheral nervous systems, including in the telencephalon. Dense α-synuclein+ pathology appears in regio inferior of the hippocampus in both Parkinson's disease and dementia
Ronika Sitapara Leang et al.
PLoS pathogens, 7(10), e1002292-e1002292 (2011-10-15)
Gammaherpesviruses such as KSHV and EBV establish lifelong persistent infections through latency in lymphocytes. These viruses have evolved several strategies to counteract the various components of the innate and adaptive immune systems. We conducted an unbiased screen using the genetically
Global Trade Item Number
| SKU | GTIN |
|---|---|
| 283541-5G | 04061826245903 |
| 283541-25G | 04061826245897 |
| 07307-25MG | 04061834034902 |
| 07307-100MG | 04061838647917 |
